Skip to Content
Merck
All Photos(1)

Documents

398039

Sigma-Aldrich

1,2-Propanediol

ACS reagent, ≥99.5%

Synonym(s):

1,2-PDO, Propylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(OH)CH2OH
CAS Number:
Molecular Weight:
76.09
Beilstein:
1340498
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

grade

ACS reagent

Quality Level

vapor density

2.62 (vs air)

vapor pressure

0.08 mmHg ( 20 °C)

Assay

≥99.5%

form

liquid

autoignition temp.

779 °F

expl. lim.

12.5 %

impurities

≤0.0005 meq/g Titr. acid
≤0.2% water

ign. residue

≤0.005%

color

APHA: ≤10

refractive index

n20/D 1.432 (lit.)

bp

187 °C (lit.)

mp

−60 °C (lit.)

density

1.036 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤1 ppm

SMILES string

CC(O)CO

InChI

1S/C3H8O2/c1-3(5)2-4/h3-5H,2H2,1H3

InChI key

DNIAPMSPPWPWGF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,2-Propanediol, a 1,2-alkanediol is an industrially important chemical. Due to its increasing demand many green methods have been proposed for its synthesis mainly using the inexpensive glycerol as the starting material. It is a raw material for the synthesis of intermediates, building blocks and polymers.

Application

1,2-Propanediol may be used in the following processes:
  • Green synthesis of propylene carbonate.
  • Production of lactic acid by green catalytic oxidation.
  • Propanal generation by catalytic dehydration.

Other Notes

The article number 398039-4X2L will be discontinued. Please order the single bottle 398039-2L which is physically identical with the same exact specifications.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

219.2 °F - closed cup

Flash Point(C)

104 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Microwave assisted process intensification of lipase catalyzed transesterification of 1, 2 propanediol with dimethyl carbonate for the green synthesis of propylene carbonate: Novelties of kinetics and mechanism of consecutive reactions.
Yadav GD, et al.
Chemical Engineering Journal, 281, 199-208 (2015)
Sustainability metrics for a fossil-and renewable-based route for 1, 2-propanediol production: A comparison.
Marinas A, et al.
Catalysis Today, 239, 31-37 (2015)
A novel Pd/C-catalyzed conversion of glucose to 1, 2-propanediol by water splitting with Zn.
Wang, J, et al.
Royal Society of Chemistry Advances, 5(63), 51435-51439 (2015)
Reactive Extraction of 1, 3-Propanediol with Aldehydes in the Presence of a Hydrophobic Acidic Ionic Liquid as a Catalyst.
Matsumoto M, et al.
Solvent Extraction Research and Development, Japan, 22(2), 209-213 (2015)
Catalytic Dehydration of 1,2-Propanediol into Propanal over Ag-Modified Silica-Alumina.
Sun D, et al.
Chemistry Letters (Jpn), 43(4), 450-452 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service