Skip to Content
Merck
All Photos(1)

Key Documents

68340

Sigma-Aldrich

Hexylene glycol

puriss., ≥99.0% (GC)

Synonym(s):

(±)-2-Methyl-2,4-pentanediol, MPD

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(OH)CH2C(CH3)2OH
CAS Number:
Molecular Weight:
118.17
Beilstein:
1098298
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

4.1 (vs air)

Quality Level

vapor pressure

0.02 mmHg ( 20 °C)

grade

puriss.

Assay

≥99.0% (GC)

form

viscous liquid

expl. lim.

7.4 %

refractive index

n20/D 1.427 (lit.)
n20/D 1.428

bp

197 °C (lit.)

mp

−40 °C (lit.)

density

0.925 g/mL at 25 °C (lit.)

SMILES string

CC(O)CC(C)(C)O

InChI

1S/C6H14O2/c1-5(7)4-6(2,3)8/h5,7-8H,4H2,1-3H3

InChI key

SVTBMSDMJJWYQN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Hexylene glycol can also be used as a solvent in the preparation of CuO doped on TiO2/silica gel photocatalyst.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

201.2 °F - closed cup

Flash Point(C)

94 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Trichloroethylene degradation by photocatalysis in annular flow and annulus fluidized bed photoreactors
Lim TH and Kim SD
Chemosphere, 54(3), 305-312 (2004)
Catherine Michaux et al.
Journal of molecular biology, 375(5), 1477-1488 (2007-12-18)
Sodium dodecyl sulfate (SDS) is a highly effective and widely used protein denaturant. We show that certain amphipathic cosolvents such as 2-methyl-2,4-pentanediol (MPD) can protect proteins from SDS denaturation, and in several cases can refold proteins from the SDS-denatured state.
Jose Antonio Cuesta-Seijo et al.
Structure (London, England : 1993), 18(9), 1210-1219 (2010-09-10)
Enzymatic reactions involving bilayer lipids occur in an environment with strict physical and topological constraints. The integral membrane enzyme PagP transfers a palmitoyl group from a phospholipid to lipid A in order to assist Escherichia coli in evading host immune
Fabio Capuano et al.
Physical chemistry chemical physics : PCCP, 13(8), 3319-3327 (2010-12-25)
The mutual diffusion coefficients for two aqueous ternary systems, both containing a protein, human serum albumin (HSA, component 1), were measured. The first system contained a neutral polymer, polyethylene glycol (PEG, component 2), and the second an "organic solvent", 2-methyl-2,4-pentanediol
Armando G Villaseñor et al.
Acta crystallographica. Section D, Biological crystallography, 68(Pt 8), 893-900 (2012-08-08)
Focused acoustic energy allows accurate and precise liquid transfer on scales from picolitre to microlitre volumes. This technology was applied in protein crystallization, successfully transferring a diverse set of proteins as well as hundreds of precipitant solutions from custom and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service