Skip to Content
Merck
All Photos(1)

Key Documents

53379

Sigma-Aldrich

FITC-CM-Dextran

average mol wt 40,000

Synonym(s):

FITC-CM-Dextran 40, Fluorescein isothiocyanate–Carboxymethyl–Dextran

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352108
NACRES:
NA.32

biological source

synthetic

Quality Level

conjugate

FITC conjugate

form

powder or crystals

mol wt

average mol wt 40,000

composition

carboxymethyl content, 3-7%

extent of labeling

0.001-0.020 mol/mol FITC glucose

technique(s)

permeability assay: suitable

fluorescence

λex 493 nm; λem 517 nm±5 nm in 0.1 M Tris pH 8.0

Looking for similar products? Visit Product Comparison Guide

Application

FITC-CM-Dextran 40, an anionic fluorescent dextran that may be used to study cell processes such as cell permeability, phagocytosis and endocytosis and to study mechanisms of biomolecular delivery. FITC-CM-Dextran 40 may be use to compare its selectivity versus neutral FITC-Dextran 40.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vincent Yi-Fong Su et al.
Stem cells (Dayton, Ohio), 37(12), 1516-1527 (2019-10-08)
Induced pluripotent stem cells (iPSCs) can reduce the severity of endotoxin-induced acute lung injury (ALI). However, the interaction between iPSCs and vascular endothelium remains unclear. In this study, we investigated the effects of iPSCs in moderating pulmonary endothelial leakage in
Rui Liu et al.
International journal of molecular medicine, 42(5), 2903-2913 (2018-08-23)
Inflammatory bowel disease (IBD) is a multifactorial inflammatory disease, and increasing evidence has demonstrated that the mechanism of the pathogenesis of IBD is associated with intestinal epithelial barrier injury. Long non‑coding RNAs (lncRNAs) are a class of transcripts >200 nucleotides
Dan Zhang et al.
Journal of cellular physiology, 233(1), 688-698 (2017-03-23)
Understanding the role and underlying regulation mechanism of autophagy in lipopolysaccharide-induced lung injury (LPS-LI) may provide potentially new pharmacological targets for treatment of acute lung injury. The aim of this study was to investigate the functional significance of autophagy in
Chichi Li et al.
Cell death & disease, 10(11), 804-804 (2019-10-28)
Adipose-derived stem cells (ADSCs) have been shown to be beneficial in some pulmonary diseases, and the paracrine effect is the major mechanism underlying ADSC-based therapy. Autophagy plays a crucial role in maintaining stem cell homeostasis and survival. However, the role
Carrier and dose effects on the pharmacokinetics of T-0128, a camptothecin analogue-carboxymethyl dextran conjugate, in non-tumor- and tumor-bearing rats.
Harada M, Murata J, Sakamura Y, et al.
J. Controlled Release, 71, 71-86 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service