Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
FITC-CM-Dextran
Synonym(s):
Fluorescein isothiocyanate–Carboxymethyl–Dextran
Products
Product Category
carbohydrates (4)
Brand
Sigma-Aldrich (4)
Biological Source
Physical Form
Chemical Composition
Conjugate
Quality Segment
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
68059
FITC-CM-Dextran,
average mol wt 4,000
Expand
View Pricing
53471
FITC-CM-Dextran,
average mol wt 70,000
Expand
View Pricing
74817
FITC-CM-Dextran,
average mol wt 150,000
Expand
View Pricing
53379
FITC-CM-Dextran,
average mol wt 40,000
Expand
View Pricing