Skip to Content
Merck
All Photos(1)

Key Documents

5100

Sigma-Aldrich

CD164 human

recombinant, expressed in E. coli, 0.5 mg protein/mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.75

biological source

human

recombinant

expressed in E. coli

description

0.05 mg of recombinant human CD164 in 20 mM Tris-HCl buffer, containing NaCl, KCl, EDTA, L-arginine, DTT and glycerol.

sterility

Filtered sterilized solution

Assay

≥90% (SDS-PAGE)

form

liquid

packaging

pkg of 50 μg

concentration

0.5 mg protein/mL

accession no.

NP_006007

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... CD164(8763)

Application

Coating a plate well (6 well plate) with this recombinant CD164 matrix protein in HSC cell specific medium at 1-10 μg/well allows for human HSC / receptor interaction studies in vitro.

Use this procedure as a guideline to determine optimal coating conditions for the culture system of choice.
1. Thaw CD164 and dilute to desired concentration using serum-free medium or PBS. The final solution should be sufficiently dilute so the volume added covers the surface evenly (1-10 μg/well, 6 well plate).
Note: Use 1 ml PBS per well in a 6-well plate.
2. Add 1 - 10 μg protein to each well and incubate at 2 to 10 °C overnight.
3. After incubation, aspirate remaining material.
4. Plates are ready for use. They may also be stored at 2-8 °C damp or air dried if sterility is maintained

Sequence

MASMTGGQQMGRGHHHHHHGNLYFQGGEFDKNTTQHPNVTTLAPISNVTSAPVTSLPLVTTPAPETCEGRNSCVSCFNVSVVNTTCFWIECKDESYCSHNSTVSDCQVGNTTDFCSVSTATPVPTANSTAKPTVQPSPSTTSKTVTTSGTTNNTVTPTSQPVRKSTFD

Preparation Note

The full-length extracellular domain of the human CD164 cDNA (24 - 162 aa) was constructed with 29 N-terminal T7/HIS-tag and expressed in E. coli as inclusion bodies. The final product was refolded using a unique “temperature shift inclusion body refolding” technology and chromatographically purified as soluble protein.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Benedikt Demmert et al.
Materials (Basel, Switzerland), 12(11) (2019-06-07)
Calcareous biominerals typically feature a hybrid nanogranular structure consisting of calcium carbonate nanograins coated with organic matrices. This nanogranular organisation has a beneficial effect on the functionality of these bioceramics. In this feasibility study, we successfully employed a flow-chemistry approach
Felipe Díaz-Soler et al.
Nanomaterials (Basel, Switzerland), 11(1) (2021-01-23)
In this work, calcium oxalate (CaOx) precursors were stabilized by poly(acrylic acid) (PAA) as an additive under in vitro crystallization assays involving the formation of pre-nucleation clusters of CaOx via a non-classical crystallization (NCC) pathway. The in vitro crystallization of
P T Ramos et al.
Materials science & engineering. C, Materials for biological applications, 103, 109742-109742 (2019-07-28)
This study aimed to develop nanocapsules containing ketoprofen using rose hip oil (Keto-NC) as oil core, and to evaluate their anti-inflammatory activity in acute and chronic ear edema models in mice. Physicochemical characterization, drug release, photostability and cytotoxicity assays were
Santosh Prasad Gupta et al.
Journal of physics. Condensed matter : an Institute of Physics journal, 32(19), 194004-194004 (2020-01-21)
We present studies on the structure of complexes of the cationic, bilayer-forming surfactant, didodecyldimethylammonium bromide (DDAB), and the anionic polyelectrolyte sodium polyacrylate (PAANa). In the presence of uncomplexed polyelectrolyte in the coexisting aqueous solution, these complexes are found to exhibit
Y Masuzawa et al.
Journal of biochemistry, 112(5), 609-615 (1992-11-01)
The peanut agglutinin (PNA)-binding site is protein-bound Gal beta 1-->3GalNAc, and is a tumor-associated carbohydrate marker expressed in many human carcinomas. PNA-binding glycoproteins isolated from KATO-III human gastric carcinoma cells were deglycosylated by trifluoromethanesulfonic acid, and rabbit antibodies against the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service