Skip to Content
Merck
All Photos(1)

Key Documents

F6892

Sigma-Aldrich

Farnesyl pyrophosphate ammonium salt

methanol:ammonia solution, ≥95% (TLC)

Synonym(s):

3,7,11-Trimethyl-2,6,10-dodecatrien-1-yl pyrophosphate ammonium salt, FPP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H37N3O7P2
CAS Number:
Molecular Weight:
433.42
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.83

Assay

≥95% (TLC)

form

methanol:ammonia solution

packaging

vial of 200 μg

storage temp.

−20°C

SMILES string

C\C(C)=C\CC\C(C)=C\CC\C(C)=C\COP(O)(=O)OP(O)(O)=O

InChI

1S/C15H28O7P2/c1-13(2)7-5-8-14(3)9-6-10-15(4)11-12-21-24(19,20)22-23(16,17)18/h7,9,11H,5-6,8,10,12H2,1-4H3,(H,19,20)(H2,16,17,18)/b14-9+,15-11+

InChI key

VWFJDQUYCIWHTN-YFVJMOTDSA-N

Gene Information

rat ... Fnta(25318)

General description

Farnesyl pyrophosphate is a 15-carbon isoprenoid synthesized from geranyl pyrophosphate (GPP) by the action of enzyme farnesyl pyrophosphate synthase (FPPS).

Application

Farnesyl pyrophosphate ammonium salt has been used:
  • as a prenylation agonist in human osteogenic sarcoma cells in collagen-based cell invasion assays
  • in the prenylation of the hepatocyte growth factor (HGF) in human umbilical vein endothelial cells (HUVECs)
  • as a substrate in prenyltransferases assay in diatom Haslea ostrearia

Biochem/physiol Actions

Farnesyl pyrophosphate (FPP) is the precursor for the biosynthesis of cholesterol, ubiquinone and dolicol. It is part of the intracellular mevalonate pathway. FPP is essential for cell survival and is used for prenylation of several low molecular mass G proteins, including Ras. Inhibition of prenylation results in loss of oncogenic potential of Ras proteins. Inhibition of prenylation may serve as therapeutic potential for management of synaptic plasticity and Alzheimer′s disease.

Physical form

Solution in methanol: 10mM aqueous NH4OH (7:3)
Actual concentration given on label

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

60.8 °F

Flash Point(C)

16 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Petra M Bleeker et al.
Proceedings of the National Academy of Sciences of the United States of America, 109(49), 20124-20129 (2012-11-22)
Tomato breeding has been tremendously efficient in increasing fruit quality and quantity but did not focus on improving herbivore resistance. The biosynthetic pathway for the production of 7-epizingiberene in a wild tomato was introduced into a cultivated greenhouse variety with
Gregory J Reynolds et al.
Plants (Basel, Switzerland), 8(7) (2019-07-25)
Systemic acquired resistance (SAR) is a mechanism through which plants may respond to initial challenge by a pathogen through activation of inducible defense responses, thereby increasing resistance to subsequent infection attempts. Fitness costs are assumed to be incurred by plants
Isoprenoid biosynthesis in the diatom Haslea ostrearia
Kampranis SC and Verret F
The New phytologist, 1-1 (2018)
Chantal Brämer et al.
Biotechnology progress, 35(4), e2812-e2812 (2019-04-02)
The natural production of patchouli oil in developing countries cannot meet the increasing demand any more. This leads to socioecological consequences, such as the use of arable land, which is actually intended for food. Hence, the world market price increased
Matrix metalloproteinase-1 contribution to sarcoma cell invasion
Garamszegi N, et al.
Journal of Cellular and Molecular Medicine, 16(6), 1331-1341 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service