Skip to Content
Merck
All Photos(1)

Key Documents

39705

Sigma-Aldrich

D-Glyceraldehyde 3-phosphate solution

8-13 mg/mL in H2O

Synonym(s):

(2R)-2-Hydroxy-3-phosphonooxypropanal, D-GAP, Triose phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H7O6P
CAS Number:
Molecular Weight:
170.06
Beilstein:
1725007
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

Assay

≥97.0% (TLC)

form

solution

concentration

8-13 mg/mL in H2O

color

colorless

storage temp.

−20°C

SMILES string

O[C@H](COP(O)(O)=O)C=O

InChI

1S/C3H7O6P/c4-1-3(5)2-9-10(6,7)8/h1,3,5H,2H2,(H2,6,7,8)/t3-/m0/s1

InChI key

LXJXRIRHZLFYRP-VKHMYHEASA-N

Application


  • Enzymatic Assay Development with d-Glyceraldehyde 3-Phosphate: Demonstrated as a crucial reagent in enzymatic assays involving glycolysis pathways, specifically in the assessment of glyceraldehyde 3-phosphate dehydrogenase activities, which play a vital role in understanding cellular energy production and metabolic disorders (Li et al., 2022).

Biochem/physiol Actions

An important substance implicated in energy metabolism. It is a key node point linking other metabolic processes or pathways. Triosephosphate metabolism can be associated with many metabolic pathways, including glycolysis, pentose phosphate pathway, gluconeogenesis and lipid metabolism, and can also be involved in maintaining hemostasis. The metabolic abnormality of triosephosphate may be tied to many pathophysiological processes, such as triosephosphate isomerase deficiency, hyperglycemia and diabetic nephropathy; review and papers cited therein.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Siripat Ngoennet et al.
Life (Basel, Switzerland), 10(3) (2020-03-19)
The halotolerant cyanobacterium, Halothece sp. PCC 7418, possesses two classes of fructose-1,6-bisphosphate aldolase (FBA): H2846 and H2847. Though class I (CI)-FBA H2846 is thought to be associated with salt tolerance, the regulatory mechanisms, molecular characteristics, and expression profiles between H2846
Derek A Franklin et al.
Scientific reports, 6, 38067-38067 (2016-12-03)
Activation of p53 in response to DNA damage is essential for tumor suppression. Although previous studies have emphasized the importance of p53-dependent cell cycle arrest and apoptosis for tumor suppression, recent studies have suggested that other areas of p53 regulation
Ruilian Yao et al.
Microbial cell factories, 10, 67-67 (2011-08-13)
Most bacteria can use various compounds as carbon sources. These carbon sources can be either co-metabolized or sequentially metabolized, where the latter phenomenon typically occurs as catabolite repression. From the practical application point of view of utilizing lignocellulose for the
Amparo C Villablanca et al.
SpringerPlus, 2(1), 214-214 (2013-06-07)
Vascular endothelium expresses both the estrogen receptors (ERs) α and β, and ERα mediates development of early atherosclerosis in male mice. This process is thought to be testosterone-dependent. We hypothesized that male murine aortic endothelium produces robust levels of estradiol
Rebeca Arroyo Hornero et al.
Communications biology, 3(1), 375-375 (2020-07-16)
Regulatory T cells (Tregs) are critical mediators of immune homeostasis. The co-stimulatory molecule CD27 is a marker of highly suppressive Tregs, although the role of the CD27-CD70 receptor-ligand interaction in Tregs is not clear. Here we show that after prolonged

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service