Skip to Content
Merck
All Photos(2)

Documents

G2267

Sigma-Aldrich

Glyceraldehyde-3-phosphate Dehydrogenase from rabbit muscle

lyophilized powder, ≥75 units/mg protein

Synonym(s):

D-Glyceraldehyde 3-phosphate:NAD+ oxidoreductase (phosphorylating), GAPDH

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

biological source

rabbit muscle

form

lyophilized powder

specific activity

≥75 units/mg protein

composition

Protein, ≥80% biuret

foreign activity

3-Phosphoglyceric phosphokinase ≤0.05%
lactic dehydrogenase, myokinase, and pyruvate kinase ≤0.01%
triosephosphate isomerase ≤0.1%

shipped in

wet ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Glyceraldehyde-3-phosphate dehydrogenase (GAPDH) is a multifunctional protein. It is basically called as a glycolytic enzyme.

Application

Glyceraldehyde-3-phosphate Dehydrogenase from rabbit muscle has been used:
  • for measurements of glyceraldehyde-3-phosphate dehydrogenase (GAPDH) activity assay
  • to generate a linear standard curve to analyse GAPDH activity in the experimental samples collected from mice
  • in anti-aggregation assays
  • in purified GAPDH studies

Biochem/physiol Actions

Glyceraldehyde-3-phosphate dehydrogenase catalyzes the conversion of glyceraldehyde-3-phosphate into D-glycerate-1,3-bisphosphate as part of the glycolysis pathway.
Glyceraldehyde-3-phosphate dehydrogenase catalyzes the conversion of glyceraldehyde-3-phosphate to 1,3-bisphosphoglycerate as part of glycolysis. It has also been shown to have roles in initiation of apoptosis, transcription activation and the shuttling of ER to Golgi vesicles<<<18,New>>>.

Unit Definition

One unit will reduce 1.0 μmole of 3-phosphoglycerate to D-glyceraldehyde 3-phosphate per min in a coupled system with 3-phosphoglyceric phosphokinase at pH 7.6 at 25 °C.

Physical form

Lyophilized powder containing citrate buffer salts

inhibitor

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Poly ADP-Ribose Polymerase Inhibition Ameliorates Hind Limb Ischemia Reperfusion Injury in a Murine Model of Type 2 Diabetes
Long C A, et al.
Annals of Surgery, 258(6) (2013)
Oligomerization of a molecular chaperone modulates its activity
Saio T, et al.
eLife, 7(6), e35731-e35731 (2018)
The C-terminal domain of Escherichia coli trigger factor represents the central module of its chaperone activity
Merz F, et al.
The Journal of Biological Chemistry, 281(42), 31963-31971 (2006)
Glyceraldehyde-3-phosphate dehydrogenase (GAPDH) aggregation causes mitochondrial dysfunction during oxidative stress-induced cell death.
Nakajima H, et al.
The Journal of Biological Chemistry, 292(11), 4727-4742 (2017)
Role of glyceraldehyde-3-phosphate dehydrogenase (GAPDH) in DNA repair
Kosova A A, et al.
The Journal of Biological Chemistry, 82(6), 643-654 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service