Skip to Content
Merck
All Photos(4)

Key Documents

D5764

Sigma-Aldrich

2,3-Diphospho-D-glyceric acid pentasodium salt

glycolysis metabolite

Synonym(s):

2,3-DPG, D-Glycerate 2,3-diphosphate sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C3H3O10P2Na5
CAS Number:
Molecular Weight:
375.95
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

biological source

synthetic

form

powder

impurities

3-phosphoglyceric acid, essentially free

color

white to off-white

solubility

water: 100 mg/mL, clear, colorless to faintly yellow

cation traces

Na: 24.5-32.5% (anhydrous)

storage temp.

−20°C

SMILES string

[Na].OC(=O)[C@@H](COP(O)(O)=O)OP(O)(O)=O

InChI

1S/C3H8O10P2.Na.H/c4-3(5)2(13-15(9,10)11)1-12-14(6,7)8;;/h2H,1H2,(H,4,5)(H2,6,7,8)(H2,9,10,11);;/t2-;;/m1../s1

InChI key

JJEMSNINYKJOBZ-YBBRRFGFSA-N

Application

D-Glycerate 2,3-diphosphate, cofactor of both phosphoglyceric acid mutase and hemoglobin, may be used as a reference compound in analysis of blood cell (erythrocyte) glycolysic cycle metabolites.

Biochem/physiol Actions

Metabolite of glycolysis within human erythrocytes produced by 2,3-diphosphoglycerate mutase in an alternative to the glycolytic pathway. Binds to hemaglobin to reduce oxygenation.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chiara Crestani et al.
Journal of veterinary internal medicine, 32(3), 1051-1059 (2018-04-11)
Optimal procedure for storage of feline blood is needed. Open-collection systems have been employed in feline medicine, thus limiting the possibility for storage. To evaluate indicators of quality of feline blood stored for 35 days at +4°C in a closed-collection
Minke A E Rab et al.
American journal of hematology, 94(5), 575-584 (2019-02-21)
In sickle cell disease (SCD), sickle hemoglobin (HbS) polymerizes upon deoxygenation, resulting in sickling of red blood cells (RBCs). These sickled RBCs have strongly reduced deformability, leading to vaso-occlusive crises and chronic hemolytic anemia. To date, there are no reliable
H Gulliksson et al.
Vox sanguinis, 112(1), 33-39 (2016-12-22)
The present general plasticizer di-2-ethylhexyl-phthalate in polyvinylchloride (PVC) blood bags is only physically dispersed in PVC and will therefore leach into blood components. The objective of this study was to perform a first preliminary red blood cell (RBC) storage evaluation
Manon Bardyn et al.
Metabolites, 10(6) (2020-06-04)
After blood donation, the red blood cells (RBCs) for transfusion are generally isolated by centrifugation and then filtrated and supplemented with additive solution. The consecutive changes of the extracellular environment participate to the occurrence of storage lesions. In this study
Ida Stangerup et al.
Transfusion, 57(8), 1905-1911 (2017-04-30)
No former studies have examined how blood donation influences physical performance in women, who due to menstruation may have a slower recovery of performance. Therefore, the aim of this study was to clarify how VO VO VO VO

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service