Skip to Content
Merck
All Photos(2)

Documents

285C

Sigma-Aldrich

Fetal Hemoglobin Kit

Synonym(s):

Modified Kleihauer-Betke procedure.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116121
NACRES:
NA.47

usage

sufficient for 50 manual assays

shelf life

Expiry date on the label.

IVD

for in vitro diagnostic use

application(s)

hematology
histology

shipped in

wet ice

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Intended for the acid elution of adult hemoglobin and the semi-quantitative determination of fetal hemaglobin in blood smears.

Kit Components Also Available Separately

Product No.
Description
SDS

  • 2851Citrate Phosphate Buffer Concentrate 100 mLSDS

  • 2852Acid Hematoxylin Solution 50 mLSDS

  • 2853Eosin B Solution, 0.1% aqueous solution 50 mLSDS

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M T Ammami et al.
Chemosphere, 125, 1-8 (2015-02-04)
Dredged harbor sediment co-contaminated by heavy metals and polycyclic aromatic hydrocarbons (PAHs) was subjected to enhanced electrokinetic treatments, using a mixture of a chelating agent (citric acid CA) and a surfactant as additives in the processing fluids. We tested various

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service