Skip to Content
Merck
All Photos(1)

Key Documents

22990-U

Supelco

Stainless Steel HPLC Fittings

nut for for 1/16 in. tubing

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
NACRES:
SB.52

material

stainless steel nut

Quality Level

packaging

pkg of 10 ea

manufacturer/tradename

VICI Valco® ZN1

fitting

nut for 1/16" tubing

Looking for similar products? Visit Product Comparison Guide

General description

The Valco stainless steel nut has a longer hex portion for easier wrench access.

Legal Information

Valco is a registered trademark of Valco Instruments Co. Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service