Skip to Content
Merck
All Photos(2)

Key Documents

R4132

Sigma-Aldrich

Reticulocyte Stain

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116121
NACRES:
NA.47

form

solution

Quality Level

shelf life

Expiry date on the label.

IVD

for in vitro diagnostic use

technique(s)

microbe id | staining: suitable

application(s)

hematology
histology

storage temp.

room temp

Application

Utilizes New Methylene Blue for the identification of reticulocytes in blood films. Reticulocyte Stain has been used in the detection of reticulocytes.

Principle

Blood and reticulocyte staining solution (New Methylene Blue) are mixed and incubated briefly at room temperature. Wedge or spun smears are made on microscope slides, air dried and evaluated under oil immersion on a light microscope. A red blood cell scoring positive would be observed containing two or more blue-stained granules.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

HDL cholesterol levels are an important factor for determining the lifespan of erythrocytes
Meurs I, et al.
Experimental Hematology, 33(11), 1309-1319 (2005)
Thiet M Vu et al.
Nature, 550(7677), 524-528 (2017-10-19)
Sphingosine-1-phosphate (S1P), a potent signalling lipid secreted by red blood cells and platelets, plays numerous biologically significant roles. However, the identity of its long-sought exporter is enigmatic. Here we show that the major facilitator superfamily transporter 2b (Mfsd2b), an orphan
Isabelle Jubinville et al.
Comparative biochemistry and physiology. Part A, Molecular & integrative physiology, 247, 110742-110742 (2020-06-15)
When successive stages of an organism's life-history overlap, conflicts and trade-offs can emerge due to competition among physiological pathways. For example, long periods of sustained locomotion in migrating birds are supported by the androgenic up-regulation of aerobic factors, such as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service