Skip to Content
Merck
All Photos(2)

Documents

14343

Sigma-Aldrich

1(S),9(R)-(−)-Bicuculline methiodide

≥95.0% (HPCE)

Synonym(s):

(5S)-5-[(6R)-6,8-Dihydro-8-oxofuro[3,4-e]-1,3-benzodioxol-6-yl]-5,6,7,8-tetrahydro-6,6-dimethyl-1,3-dioxolo[4,5-g]isoquinolinium iodide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H20INO6
CAS Number:
Molecular Weight:
509.29
Beilstein:
5419649
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥95.0% (HPCE)

form

powder

optical activity

[α]/D -117±7°, c = 1 in chloroform

solubility

H2O: 10 mg/mL, almost clear, greenish-yellow

storage temp.

2-8°C

SMILES string

[I-].[H][C@]1(OC(=O)c2c3OCOc3ccc12)[C@]4([H])c5cc6OCOc6cc5CC[N+]4(C)C

InChI

1S/C21H20NO6.HI/c1-22(2)6-5-11-7-15-16(26-9-25-15)8-13(11)18(22)19-12-3-4-14-20(27-10-24-14)17(12)21(23)28-19;/h3-4,7-8,18-19H,5-6,9-10H2,1-2H3;1H/q+1;/p-1/t18-,19+;/m0./s1

InChI key

HKJKCPKPSSVUHY-GRTNUQQKSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Bicuculline is a phthalide isoquinoline alkaloid, which is extracted from Dicentra cucullaria. It acts as an acetylcholinesterase inhibitor. Bicuculline inhibits slow afterhyperpolarization (AHP).

Application

1(S),9(R)-(-)-Bicuculline methiodide has been used as a gamma-aminobutyric acid (GABA) antagonist to block inhibition in the semicoronal slices. It has also been used for the preparation of organotypic hippocampal slice cultures (OHSCs).

Biochem/physiol Actions

(-)-Bicuculline methiodide is a GABAA receptor antagonist, which acts as a competitive inhibitor of GABA ligand binding to the receptor.

Features and Benefits

This compound is a featured product for Neuroscience research. Click here to discover more featured Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the GABAA Receptors and Potassium Channels pages of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Preparation Note

prepared from (+)-bicuculline

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Bicuculline block of small-conductance calcium-activated potassium channels
Khawaled R, et al.
Pfluegers Archiv, 438(3), 314-321 (1999)
Dong Hyun Kim et al.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology, 37(2), 422-433 (2011-09-09)
Memory consolidation is the process by which acquired information is converted to something concrete to be retrieved later. Here we examined a potential role for brain-derived neurotrophic factor (BDNF) in mediating the enhanced memory consolidation induced by the GABA(A) receptor
Electrophysiological characterization of networks and single cells in the hippocampal region of a transgenic rat model of Alzheimer?s disease
Heggland I, et al.
eNeuro, 6(1) (2019)
Charles Capaday et al.
The Journal of physiology, 589(Pt 10), 2515-2528 (2011-04-14)
Motor cortical points are linked by intrinsic horizontal connections having a recurrent network topology. However, it is not known whether neural activity can propagate over the area covered by these intrinsic connections and whether there are spatial anisotropies of synaptic
Shoko Iida et al.
Biochemical and biophysical research communications, 501(1), 152-157 (2018-05-04)
The balance between glutamate-mediated excitation and GABA-mediated inhibition is critical to cortical functioning. However, the contribution of network structure consisting of the both neurons to cortical functioning has not been elucidated. We aimed to evaluate the relationship between the network

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service