Skip to Content
Merck
All Photos(3)

Key Documents

445460

Sigma-Aldrich

HATU

97%, for peptide synthesis

Synonym(s):

1-[Bis(dimethylamino)methylene]-1H-1,2,3-triazolo[4,5-b]pyridinium 3-oxid hexafluorophosphate, N-[(Dimethylamino)-1H-1,2,3-triazolo-[4,5-b]pyridin-1-ylmethylene]-N-methylmethanaminium hexafluorophosphate N-oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H15F6N6OP
CAS Number:
Molecular Weight:
380.23
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Product Name

HATU, 97%

Assay

97%

form

solid

reaction suitability

reaction type: Coupling Reactions

mp

183-185 °C (lit.)

application(s)

peptide synthesis

functional group

amine

storage temp.

2-8°C

SMILES string

F[P-](F)(F)(F)(F)F.CN(C)[C+](N(C)C)n1n[n+]([O-])c2ncccc12

InChI

1S/C10H15N6O.F6P/c1-13(2)10(14(3)4)15-8-6-5-7-11-9(8)16(17)12-15;1-7(2,3,4,5)6/h5-7H,1-4H3;/q+1;-1

InChI key

FKBFHOSFPRWJNV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Peptide coupling reagent.[1][2][3]
Preparation of N-arylsulfonamide-linked peptides by solid-phase synthesis.[4]
Reagent for:
Synthesis of Aurora A kinase inhibitors
HPLC assay to determine D- and L- acid enantiomers in human plasma
Amide bond formation reactions

Catalyst for:
Selective acylation
Selecocyclization-oxidation deselenation sequence

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1A

Supplementary Hazards

Storage Class Code

4.1A - Other explosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yi He et al.
Organic letters, 8(12), 2483-2485 (2006-06-02)
N-Acylsulfonamide safety-catch linkers are versatile tools in solid-phase organic synthesis because of their stability. This stability necessitates linker activation prior to compound cleavage. Here, we demonstrate that the N-acylsulfonamide group can react with a pi-allyl palladium complex and that these
Lett. Pept. Sci., 9, 119-123 (2003)
J. Prakt. Chem./Chem.-Ztg., 340, 581-583 (1998)
Pept., Proceedings of the European Peptide Symposium., ed. U. Ragnarrson, 27, 272-273 (2002)
Mazen Jamous et al.
Nuclear medicine and biology, 41(6), 464-470 (2014-05-02)
The gastrin-releasing peptide receptor (GRPR) was shown to be expressed with high density on several types of cancers. Radiolabeled peptides for imaging and targeted radionuclide therapy have been developed. In this study, we evaluated the potential of statine-based bombesin antagonists

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service