Skip to Content
Merck
All Photos(2)

Key Documents

107689

Sigma-Aldrich

Hexadimethrine bromide

≥95%

Synonym(s):

1,5-Dimethyl-1,5-diazaundecamethylene polymethobromide, Polybrene

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41106502
NACRES:
NA.26

Assay

≥95%

storage temp.

2-8°C

SMILES string

BrCCCBr.N(CCCCCCN(C)C)(C)C

InChI

1S/C10H24N2.C3H6Br2/c1-11(2)9-7-5-6-8-10-12(3)4;4-2-1-3-5/h5-10H2,1-4H3;1-3H2

InChI key

KZKAYEGOIJEWQB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Adaptive and Ultrahigh-Affinity Recognition in Water by Sulfated Conjugated Corral[5]arene.: Research on Hexadimethrine bromide′s interaction with sulfated corral[5]arene under aqueous conditions, exploring its high-affinity binding properties which could be crucial for developing new biochemical sensors (Wang et al., 2024).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Montalescot et al.
Circulation, 82(5), 1754-1764 (1990-11-01)
Protamine reversal of heparin anticoagulation in patients is occasionally associated with life-threatening acute pulmonary hypertension. In a sheep model, we evaluated the effect on this adverse cardiopulmonary reaction of modifying the type of heparin (low molecular weight heparin compared with
Noam Auslander et al.
Molecular systems biology, 13(12), 956-956 (2017-12-03)
Metabolic alterations play an important role in cancer and yet, few metabolic cancer driver genes are known. Here we perform a combined genomic and metabolic modeling analysis searching for metabolic drivers of colorectal cancer. Our analysis predicts FUT9, which catalyzes
Yujie Zhang et al.
Scientific reports, 7, 41173-41173 (2017-01-24)
Excessive deposition of type I collagen causes fibrotic diseases. Binding of La ribonucleoprotein domain family, member 6 (LARP6) to collagen mRNAs regulates their translation and is necessary for high type I collagen expression. Here we show that mTORC1 phosphorylates LARP6
Yuji Kondo et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(12), 4714-4719 (2013-03-09)
Although endogenous ligands for Toll-like receptor (TLR)4-myeloid differentiation factor 2 (MD2) have not been well-understood, we here report that a globo-series glycosphingolipid, globotetraosylceramide (Gb4), attenuates the toxicity of lipopolysaccharides (LPSs) by binding to TLR4-MD-2. Because α1,4-galactosyltransferase (A4galt)-deficient mice lacking globo-series
Abdulrahman Jama et al.
The Journal of physiology, 597(3), 889-901 (2018-12-05)
Lipin1 is critical for skeletal muscle development. Lipin1 regulates MyoD and myocyte-specific enhancer factor 2C (MEF2c) expression via the protein kinase C (PKC)/histone deacetylase 5-mediated pathway. Inhibition of PKCμ activity suppresses myoblast differentiation by inhibiting MyoD and MEF2c expression. Our

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service