Skip to Content
Merck
All Photos(1)

Documents

09833

Sigma-Aldrich

Ammonium citrate dibasic

BioXtra, ≥99.0% (T)

Synonym(s):

Ammonium hydrogencitrate, Citric acid ammonium salt, Diammonium hydrogen citrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC(CO2H)(CH2CO2NH4)2
CAS Number:
Molecular Weight:
226.18
Beilstein:
4925760
EC Number:
MDL number:
UNSPSC Code:
12161703
PubChem Substance ID:
NACRES:
NA.26

vapor density

1.8 (vs air)

product line

BioXtra

Assay

≥99.0% (T)

form

powder or crystals

impurities

insoluble matter, passes filter test

pH

4.0-5.5 (25 °C, 1 M in H2O)

solubility

H2O: 1 M at 20 °C, clear, colorless

anion traces

chloride (Cl-): ≤5 mg/kg
phosphate (PO43-): ≤5 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Al: ≤5 mg/kg
As: ≤0.1 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

λ

1 M in H2O

UV absorption

λ: 260 nm Amax: ≤0.05
λ: 280 nm Amax: ≤0.03

SMILES string

N.N.OC(=O)CC(O)(CC(O)=O)C(O)=O

InChI

1S/C6H8O7.2H3N/c7-3(8)1-6(13,5(11)12)2-4(9)10;;/h13H,1-2H2,(H,7,8)(H,9,10)(H,11,12);2*1H3

InChI key

YXVFQADLFFNVDS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Analytical procedure for the simultaneous voltammetric determination of trace metals in food and environmental matrices. Critical comparison with atomic absorption spectroscopic measurements.: This paper presents an analytical procedure utilizing ammonium citrate dibasic for the simultaneous voltammetric determination of trace metals in food and environmental samples. The study demonstrates the method′s efficacy compared to traditional atomic absorption spectroscopy (Melucci et al., 2007).

  • Overlapping voltammetric peaks--an analytical procedure for simultaneous determination of trace metals. Application to food and environmental matrices.: The study details an analytical method using ammonium citrate dibasic to resolve overlapping voltammetric peaks, allowing for the simultaneous determination of multiple trace metals in complex matrices. This technique is beneficial for environmental monitoring and food safety assessments (Locatelli, 2005).

  • Correlation of biological value of feed phosphates with their solubility in water, dilute hydrogen chloride, dilute citric acid, and neutral ammonium citrate.: This research investigates the solubility of feed phosphates in various solvents, including ammonium citrate dibasic, to determine their biological value. The findings help optimize animal feed formulations for better nutritional outcomes (Sullivan et al., 1992).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lorina Gjonaj et al.
Organic & biomolecular chemistry, 13(21), 6059-6065 (2015-05-08)
A new method for the selective chemical modification of DNA at cytosine nucleobases using alkoxy- and benzyloxyamines is presented. It is shown that in particular benzyloxyamines are effective DNA modifying agents, giving rise to almost exclusive formation of the mono
S Vollmer et al.
Organic & biomolecular chemistry, 13(20), 5734-5742 (2015-04-23)
Triplexes with a gap in the purine strand have been shown to bind adenosine or guanosine derivatives through a combination of Watson-Crick and Hoogsteen base pairing. Rigidifying the binding site should be advantageous for affinity. Here we report that clamps
Shi-Jun Dong et al.
The international journal of biochemistry & cell biology, 68, 33-41 (2015-08-19)
During the industrial bioethanol fermentation, Saccharomyces cerevisiae cells are often stressed by bacterial contaminants, especially lactic acid bacteria. Generally, lactic acid bacteria contamination can inhibit S. cerevisiae cell growth through secreting lactic acid and competing with yeast cells for micronutrients
Ewa Radzikowska et al.
Organic & biomolecular chemistry, 13(1), 269-276 (2014-11-05)
Chimeric oligonucleotides containing phosphodiester and phosphorothioate linkages have been obtained using the solid phase synthesis. The oligonucleotide parts possessing natural internucleotide phosphate bonds were assembled using commercially available nucleoside 3'-O-(2-cyanoethyl-N,N-diisopropylamino)phosphoramidites 7 whereas the phosphorothioate segment was built using nucleoside 3'-O-(2-thio-1,3,2-oxathiaphospholanes)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service