Skip to Content
Merck
All Photos(1)

Key Documents

09817

Sigma-Aldrich

Collodion solution

for microscopy, 2% in amyl acetate

Synonym(s):

Cellulose nitrate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12171500
NACRES:
NA.47
Pricing and availability is not currently available.

biological source

synthetic

grade

for microscopy

form

liquid

concentration

2% in amyl acetate

color

colorless

refractive index

n/D 1.390-1.410

application(s)

hematology
histology

storage temp.

room temp

SMILES string

[N+](=O)([O-])OC3OC(C(C(C3O[N+](=O)[O-])O[N+](=O)[O-])OC4OC(C(C(C4O[N+](=O)[O-])O[N+](=O)[O-])O[N+](=O)[O-])CO[N+](=O)[O-])CO[N+](=O)[O-].O1C(C(C(C(C1CO)O)O)O)OC2C(OC(C(C2O)O)O)CO

InChI

1S/C12H14N8O27.C12H22O11/c21-13(22)37-1-3-5(7(43-16(27)28)10(46-19(33)34)12(40-3)47-20(35)36)41-11-9(45-18(31)32)8(44-17(29)30)6(42-15(25)26)4(39-11)2-38-14(23)24;13-1-3-5(15)6(16)9(19)12(22-3)23-10-4(2-14)21-11(20)8(18)7(10)17/h3-12H,1-2H2;3-20H,1-2H2

InChI key

OWWGYSXLINVAES-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Collodion solution (celluloase nitrate) is use to prepare nitrocellulose films (collodion film) on microscope slides.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

100.4 °F

Flash Point(C)

38 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

V P Shinkarev et al.
FEBS letters, 452(3), 223-227 (1999-07-01)
Lipid-impregnated collodion (nitrocellulose) films have been frequently used as a fusion substrate in the measurement and analysis of electrogenic activity in biological membranes and proteoliposomes. While the method of fusion of biological membranes or proteoliposomes with such films has found
Xiaoli Zhang et al.
Analytical chemistry, 79(3), 1256-1261 (2007-02-01)
An electrochemical method for quantitative determination of enzyme activity in single cells was developed by scanning a microelectrode (ME) over a nitrocellulose film-covered microreactor with micropores by means of a scanning electrochemical microscope (SECM). Peroxidase (PO) in neutrophils was chosen
Gianluca Picariello et al.
Analytical chemistry, 82(13), 5783-5791 (2010-06-11)
The potential of matrix-assisted laser desorption ionization time-of-flight (MALDI-TOF) mass spectrometry (MS) for the analysis of intact triacylglycerols (TAGs) is generally limited by the extensive in-source prompt fragmentation. The sequential deposition of matrix and TAGs over the stainless steel target
Marina Cretich et al.
Analytical biochemistry, 397(1), 84-88 (2009-10-06)
We report on the modification of a nitrocellulose film with copoly(DMA-NAS-MAPS), a tercopolymer based on N,N-dimethylacrylamide (DMA), N-acryloyloxysuccinimide (NAS), and 3-(trimethoxysilyl)propyl-methacrylate (MAPS). The chains of this polymer, interacting with nitrocellulose fibers, introduce active ester functionalities that promote the covalent binding
Gina E Fridley et al.
Lab on a chip, 12(21), 4321-4327 (2012-09-11)
Novel methods are demonstrated that enable controlled spatial and temporal rehydration of dried reagents in a porous matrix. These methods can be used in paper-based microfluidic assays to define reagent concentrations over time at zones downstream for improved performance, and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service