Skip to Content
Merck
All Photos(1)

Key Documents

09986

Sigma-Aldrich

Collodion solution

4-8% in ethanol/diethyl ether

Synonym(s):

Cellulose nitrate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.22

Quality Level

concentration

4-8% in ethanol/diethyl ether

InChI

1S/C12H14N8O27.C12H22O11/c21-13(22)37-1-3-5(7(43-16(27)28)10(46-19(33)34)12(40-3)47-20(35)36)41-11-9(45-18(31)32)8(44-17(29)30)6(42-15(25)26)4(39-11)2-38-14(23)24;13-1-3-5(15)6(16)9(19)12(22-3)23-10-4(2-14)21-11(20)8(18)7(10)17/h3-12H,1-2H2;3-20H,1-2H2

InChI key

OWWGYSXLINVAES-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Collodion solution is used:
  • In the nitrocellulose treatment of working electrode in the tissue implanted biological oxygen sensor to prevent electrode poisoning.
  • As a stabilizer in synthesizing silver nanoparticles (AgNPs) to prepare silver-based conductive printing ink.
  • As a binder in the preparation of titanium powder by metallothermic reduction method.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 1 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-61.6 °F - closed cup

Flash Point(C)

-52 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Titanium powder production by preform reduction process (PRP).
Okabe TH, et al.
J. Alloy Compounds, 364(1-2), 156-163 (2004)
Real-time monitoring of brain tissue oxygen using a miniaturized biotelemetric device implanted in freely moving rats.
Bazzu G, et al.
Analytical Chemistry, 81(6), 2235-2241 (2009)
Nitrocellulose-stabilized silver nanoparticles as low conversion temperature precursors useful for inkjet printed electronics.
Nguyen BT, et al.
Journal of Materials Chemistry, 17(17), 1725-1730 (2007)
L S Selvakumar et al.
Analytica chimica acta, 722, 107-113 (2012-03-27)
In this article, we describe a dipstick based immunochemiluminescence (immuno-CL) biosensor for the detection of vitamin B(12) in energy drinks. The method is a direct competitive type format involving the immobilization of vitamin B(12) antibody on nitrocellulose membrane (NC) followed
Nelia Cordeiro et al.
Methods in molecular biology (Clifton, N.J.), 979, 43-49 (2013-02-12)
The prototypical death receptor Fas (also known as CD95 or Apo-1) plays an essential role in the maintenance of lymphocyte homeostasis. Propagation of cell death through Fas relies on the formation of a multiprotein complex at the receptor level known

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service