Skip to Content
Merck
All Photos(3)

Key Documents

N4882

Sigma-Aldrich

Nickel(II) sulfate hexahydrate

ReagentPlus®, powder or crystals

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NiSO4 · 6H2O
CAS Number:
Molecular Weight:
262.85
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
Assay:
98.0-102.0% (EDTA titration)
form:
powder or crystals

vapor density

2.07 (vs air)

Quality Level

product line

ReagentPlus®

Assay

98.0-102.0% (EDTA titration)

form

powder or crystals

cation traces

Fe: ≤0.001%

SMILES string

O.O.O.O.O.O.[Ni++].[O-]S([O-])(=O)=O

InChI

1S/Ni.H2O4S.6H2O/c;1-5(2,3)4;;;;;;/h;(H2,1,2,3,4);6*1H2/q+2;;;;;;;/p-2

InChI key

RRIWRJBSCGCBID-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Nickel(II) sulfate hexahydrate (NiSO4 · 6H2O) is commonly used as a nickel precursor and catalyst in the various organic synthesis.

Application

Nickel(II) sulfate hexahydrate can be used:
  • As a catalyst for the synthesis of α-aminophosphonates by a condensation reaction of aromatic aldehydes, primary amines, and diethylphosphite.
  • In the synthesis of nickel (II) hexamethylenetetramine complex [Ni(HMTA)2 (NCS)2(H2O)2]H2O with thiocyanate coligand.
  • As a nickel source in the synthesis of nanosized nickel phosphides by the solvothermal route using red phosphorus as the P-3 precursor.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1A Inhalation - Muta. 2 - Repr. 1B - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 1 Inhalation

Target Organs

Respiratory Tract

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eun A Choi et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 39(25), 4945-4958 (2019-04-14)
Decision-making often involves motivational conflict because of the competing demands of approach and avoidance for a common resource: behavior. This conflict must be resolved as a necessary precursor for adaptive behavior. Here we show a role for the paraventricular thalamus
Asymmetric induction by retgersite, nickel sulfate hexahydrate, in conjunction with asymmetric autocatalysis.
Matsumoto A, et al.
New. J. Chem., 39(9), 6742-6745 (2015)
Structure of monoclinic nickel (II) sulfate hexahydrate.
Gerkin RE and Reppart WJ.
Acta Crystallographica Section C, Crystal Structure Communications, 44(8), 1486-1488 (1988)
Alison R Weiss et al.
Scientific reports, 10(1), 6970-6970 (2020-04-26)
Recently, AAV2.retro, a new capsid variant capable of efficient retrograde transport in brain, was generated in mice using a directed evolution approach. However, it remains unclear to what degree transport will be recapitulated in the substantially larger and more complex
Polymorphism of nickel sulfate hexahydrate.
Angel RJ and Finger LW.
Acta Crystallographica Section C, Crystal Structure Communications, 44(11), 1869-1873 (1988)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service