Skip to Content
Merck
All Photos(1)

Documents

97876

Sigma-Aldrich

Activated Charcoal Norit®

greener alternative

Norit® CA1, wood, chemically activated, powder

Synonym(s):

Charcoal activated Norit®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C
CAS Number:
Molecular Weight:
12.01
Beilstein:
4360473
EC Number:
MDL number:
UNSPSC Code:
23201100
PubChem Substance ID:
NACRES:
SB.52

biological source

wood

form

powder

quality

chemically activated

manufacturer/tradename

Norit® CA1

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

technique(s)

ion chromatography: suitable

ign. residue

≤4.0%

loss

≤16% loss on drying

greener alternative category

SMILES string

[C]

InChI

1S/C

InChI key

OKTJSMMVPCPJKN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Activated carbon (AC) is a three dimensional highly microporous structure with high surface area. AC can be prepared from various carbon precursors, such as lignite, peat, wood, coal etc. AC powder is popularly used in adsorption or at catalyst support. The adsorption capability of activated carbon is determined by its pore size distribution, purity and degree of carbonization, surface chemistry. Method of activation also influences the final properties of the product. Activated carbon prepared from lignite are hard textured with small micropore volume and high pore size distribution.4 It is used as a carrier for catalysts and possess good filtering properties.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Click here for more details.

Application

Activated carbons may be used in:
  • Separating gas mixtures, such as; hydrogen-hydrocarbons,
  • Gas drying;
  • Selective gas adsorption
  • Gas storage
Activated charcoal Norit may be used in recovery of lithium for recycling lithium- ion batteries. It may be used to study the adsorption isotherm of 2,4,6-trichlorophenol (TCP). 6 Activated carbon may be used for ultrapurification of water, such as removal of anti-anxiety drug diazepam (valium) from spiked wastewater.

Legal Information

Norit is a registered trademark of Norit N.V.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Sol. 2

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Adsorption of 2, 4, 6-trichlorophenol (TCP) onto activated carbon.
Krishnaiah D, et al.
Journal of King Saud University - Science, 25(3), 251-255 (2013)
Vartika Mishra et al.
Nature communications, 11(1), 5579-5579 (2020-11-06)
Cell-to-cell communications are critical determinants of pathophysiological phenotypes, but methodologies for their systematic elucidation are lacking. Herein, we propose an approach for the Systematic Elucidation and Assessment of Regulatory Cell-to-cell Interaction Networks (SEARCHIN) to identify ligand-mediated interactions between distinct cellular compartments.
Diazepam stability in wastewater and removal by advanced membranes technology, activated carbon and micelle-clay complex
Bufo SA, et al.
Desalination and Water Treatment (2015)
Activated carbon for gas separation and storage
Sircar S, et al.
Carbon, 34(1), 1-12 (1996)
Lithium recovery from aqueous solution by sorption/desorption
Lemaire J, et al.
Hydrometallurgy, 143, 1-11 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service