Skip to Content
Merck
All Photos(1)

Documents

29204

Sigma-Aldrich

Activated Charcoal Norit®

greener alternative

Norit® RB3, for gas purification, steam activated, rod

Synonym(s):

Charcoal activated Norit®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C
CAS Number:
Molecular Weight:
12.01
Beilstein:
4360473
EC Number:
MDL number:
UNSPSC Code:
23201100
PubChem Substance ID:
NACRES:
SB.52

grade

for gas purification

description

butane adsorption 18 - 23% (p/p0 = 1)

form

rod

quality

steam activated

manufacturer/tradename

Norit® RB3

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

diam.

~3 mm

loss

≤5% loss on drying

diameter

~3 mm

greener alternative category

SMILES string

[C]

InChI

1S/C

InChI key

OKTJSMMVPCPJKN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Click here for more details.

Application

Activated Charcoal Norit® (Norit RB3) can be used in the separation of methane and nitrogen. It can be incorporated with MIL-101 for the formation of metal-organic frameworks (MOFs) for potential usage in hydrogen storage.

Legal Information

Norit is a registered trademark of Norit N.V.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adsorption equilibria and kinetics of methane+ nitrogen mixtures on the activated carbon Norit RB3
Rufford TE, et al.
Industrial & Engineering Chemistry Research, 52(39), 14270-14281 (2013)
Doping activated carbon incorporated composite MIL-101 using lithium: impact on hydrogen uptake
Prabhakaran PK and Deschamps J
Journal of Material Chemistry A, 3(13), 7014-7021 (2015)
Adsorption of volatile organic compounds by means of activated carbon fibre-based monoliths.
Fuertes AB, et al.
Carbon, 41(1), 87-96 (2003)
Co-adsorption of n-butane/water vapour mixtures on activated carbon fibre-based monoliths.
Marban G and Fuertes AB
Carbon, 42(1), 71-81 (2004)
Adsorption of carbon dioxide using impregnated activated carbon promoted by Zinc.
Somy A, et al.
International Journal of Greenhouse Gas Control, 3(3), 249-254 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service