Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Activated Charcoal Norit
®
Synonym(s):
Charcoal activated Norit
®
Empirical Formula (Hill Notation):
C
CAS Number:
7440-44-0
Molecular Weight:
12.01
EC Number:
231-153-3
Products
Product Category
adsorbents (1)
Brand
Sigma-Aldrich (2)
Sustainability
Biological Source
Feature
Formula Weight
Manufacturer
Physical Form
Tissue
Diameter (mm)
Technique
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
22874
Activated Charcoal Norit
®
,
Norit
®
PK 1-3, from peat, steam activated, granular
Expand
View Pricing
37771
Activated Charcoal Norit
®
,
Norit
®
GAC 1240W, from coal, steam activated, granular
Expand
View Pricing
93067
Activated Charcoal Norit
®
,
Norit
®
SX2, powder, from peat, multi-purpose activated charcoal, steam activated and acid washed
Expand
View Pricing
29204
Activated Charcoal Norit
®
,
Norit
®
RB3, steam activated, rod
Expand
View Pricing
97876
Activated Charcoal Norit
®
,
Norit
®
CA1, wood, chemically activated, powder
Expand
View Pricing