Skip to Content
Merck
All Photos(1)

Key Documents

71800

Sigma-Aldrich

Sodium oxalate

puriss. p.a., ACS reagent, ≥99.5% (RT)

Synonym(s):

Ethanedioic acid sodium salt, Oxalic acid disodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaOOCCOONa
CAS Number:
Molecular Weight:
134.00
Beilstein:
3631622
EC Number:
MDL number:
UNSPSC Code:
12352300
eCl@ss:
39021701
PubChem Substance ID:
NACRES:
NB.24
Assay:
≥99.5% (RT)
form:
powder or crystals

grade

ACS reagent
puriss. p.a.

Quality Level

Assay

≥99.5% (RT)

form

powder or crystals

impurities

≤0.002% ammonium (NH4)

loss

≤0.01% loss on drying, 105 °C

pH

7-8.5 (25 °C, 2.5% in H2O)

anion traces

chloride (Cl-): ≤20 mg/kg
sulfate (SO42-): ≤20 mg/kg

cation traces

Ca: ≤100 mg/kg
Cd: ≤10 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤10 mg/kg
K: ≤50 mg/kg
Mg: ≤10 mg/kg
Mn: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

SMILES string

[Na+].[Na+].[O-]C(=O)C([O-])=O

InChI

1S/C2H2O4.2Na/c3-1(4)2(5)6;;/h(H,3,4)(H,5,6);;/q;2*+1/p-2

InChI key

ZNCPFRVNHGOPAG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Sodium oxalate is a sodium salt of oxalic acid consisting of sodium and oxalate ions. It is used as a reducing agent in chemical synthesis. Additionally, it also acts as a surface modifier and a desulfurizing agent.

Application

Sodium oxalate can be used as a stabilizer in the determination of L-ascorbic acid in vitamin C products by the ultraviolet spectrophotometric method. It is also used in the synthesis of copper oxalate nanoparticles by electrolysis of copper plate as anode.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F Debela et al.
Journal of environmental management, 116, 156-162 (2013-01-15)
Pyromorphite (PY) and some zinc phosphates (Zn-P) are very sparingly soluble minerals and hence can immobilize Pb and Zn in contaminated soils. However, mechanisms leading to the poor efficiency of PY and Zn-P formation in contaminated soils amended with P
Lars Poulsen et al.
PloS one, 7(12), e50596-e50596 (2012-12-20)
Acid formation in Aspergillus niger is known to be subjected to tight regulation, and the acid production profiles are fine-tuned to respond to the ambient pH. Based on transcriptome data, putative trans-acting pH responding transcription factors were listed and through
Jiří Jan et al.
Water research, 47(2), 547-557 (2012-12-12)
Correct identification of P forms together with their main Fe and Al binding partners in non-calcareous sediments is of crucial importance for evaluation of P cycling in water bodies. In this paper, we assess extraction methods frequently used for this
Lu Gao et al.
International journal of phytoremediation, 15(4), 307-319 (2013-03-16)
Phytolacca americana L. can accumulate large amounts of heavy metals in its aerial tissues, especially cadmium (Cd) and manganese (Mn). It has great potential for use in phytoextraction of metals from multi-metal-contaminated soils. This study was conducted to further investigate
Taiying Zhang et al.
Carbohydrate polymers, 92(1), 334-344 (2012-12-12)
Dilute oxalic acid pretreatment was applied to maple wood to improve compatibility with downstream operations, and its performance in pretreatment and subsequent enzymatic hydrolysis was compared to results for hydrothermal and dilute hydrochloric and sulfuric acid pretreatments. The highest total

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service