379735
Sodium oxalate
≥99.99% trace metals basis
Synonym(s):
Ethanedioic acid sodium salt, Oxalic acid disodium salt
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Linear Formula:
NaOOCCOONa
CAS Number:
Molecular Weight:
134.00
Beilstein:
3631622
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
≥99.99% trace metals basis
form:
powder
Recommended Products
Quality Level
Assay
≥99.99% trace metals basis
form
powder
impurities
≤100.0 ppm Trace Metal Analysis
SMILES string
[Na+].[Na+].[O-]C(=O)C([O-])=O
InChI
1S/C2H2O4.2Na/c3-1(4)2(5)6;;/h(H,3,4)(H,5,6);;/q;2*+1/p-2
InChI key
ZNCPFRVNHGOPAG-UHFFFAOYSA-L
Looking for similar products? Visit Product Comparison Guide
Application
- Analytical procedures and methods validation for oxalate content estimation: Validates methods for estimating oxalate content, using sodium oxalate as a reducing agent in various analytical procedures (D Karamad et al., 2019).
- High-efficiency cathode sodium compensation for sodium-ion batteries: Explores the use of sodium oxalate in enhancing the performance of sodium-ion batteries (YB Niu et al., 2020).
- Solubility of sodium oxalate in Concentrated electrolyte solutions: Discusses the solubility characteristics of sodium oxalate in various electrolyte solutions at different temperatures (G Hefter, A Tromans, PM May, 2018).
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Oral
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Lars Poulsen et al.
PloS one, 7(12), e50596-e50596 (2012-12-20)
Acid formation in Aspergillus niger is known to be subjected to tight regulation, and the acid production profiles are fine-tuned to respond to the ambient pH. Based on transcriptome data, putative trans-acting pH responding transcription factors were listed and through
F Debela et al.
Journal of environmental management, 116, 156-162 (2013-01-15)
Pyromorphite (PY) and some zinc phosphates (Zn-P) are very sparingly soluble minerals and hence can immobilize Pb and Zn in contaminated soils. However, mechanisms leading to the poor efficiency of PY and Zn-P formation in contaminated soils amended with P
Bhagawatilal Jagetiya et al.
Chemosphere, 91(5), 692-696 (2012-12-27)
A greenhouse experiment was set up to investigate the ability of citric acid (CA), oxalic acid (OA), nitrilotriacetic acid (NTA) and EDTA for phytoremediation of uranium tailings by Indian mustard [Brassica juncea (L.) Czern. et Coss]. Uranium tailings were collected
Jiří Jan et al.
Water research, 47(2), 547-557 (2012-12-12)
Correct identification of P forms together with their main Fe and Al binding partners in non-calcareous sediments is of crucial importance for evaluation of P cycling in water bodies. In this paper, we assess extraction methods frequently used for this
Hong-Joo Lee et al.
Bioresource technology, 130, 211-217 (2013-01-12)
The present study investigated the feasibility of the recovery and reuse oxalic acid in a multistage process for the pretreatment of a lignocellulosic biomass. Electrodialysis (ED), an electrochemical process using ion exchange membranes, was used to recover and reuse oxalic
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service