Skip to Content
Merck
All Photos(2)

Documents

223891

Sigma-Aldrich

Bismuth(III) oxide

ReagentPlus®, powder, 10 μm, 99.9% trace metals basis

Synonym(s):

Dibismuth trioxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Bi2O3
CAS Number:
Molecular Weight:
465.96
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

product line

ReagentPlus®

Assay

99.9% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: bismuth

particle size

10 μm

SMILES string

O=[Bi]O[Bi]=O

InChI

1S/2Bi.3O

InChI key

WMWLMWRWZQELOS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Bismuth (III) oxide is a yellow, monoclinic crystalline powder. It is insoluble in water and hydroxide solutions but dissolves in acids to form bismuth (III) salts. It can be prepared by heating bismuth in air or by heating hydroxides, carbonates or nitrates of bismuth.

Application

General applications of bismuth (III) oxide are:production of optical glasses with high indices of refraction, precursor in the production of other bismuth compounds.
Bismuth (III) oxide is suitable for dental materials to render them opaque to X-rays.
It is used for the synthesis of pure BiFeO3 perovskite nanoparticles, lead free piezoelectric 0.96(Bi0.5Na0.5)TiO3 - 0.04 BaTiO3.

Features and Benefits

Preparation of new stabilized, oxide ion-conducting, bismuth vanadate phases by a microwave assisted method, from V2O5, Bi2O3 and other solid oxides, was reported. These ceramics show promise in solid oxide fuel cells, water-vapor electrolyzers and oxygen sensors.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

nwg

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Direct mechanosynthesis of pure BiFeO 3 perovskite nanoparticles: reaction mechanism.
Perejon A, et al.
Journal of Material Chemistry C, 1(22), 3551-3562 (2013)
Josette Camilleri et al.
PloS one, 15(11), e0240634-e0240634 (2020-11-12)
Bismuth(III) oxide is included as a radio-opacifier in dental materials, including hydraulic silicate cements, the material of choice for several endodontic procedures. It has been implicated in tooth discoloration after contact with endodontic irrigants, in particular NaOCl solution, To date
Atsushi Satsuma et al.
Sensors (Basel, Switzerland), 11(2), 2155-2165 (2012-02-10)
Mixed potential sensors were fabriated using yttria-stabilized zirconia (YSZ) as a solid electrolyte and a mixture of Au and various metal oxides as a sensing electrode. The effects of calcination temperature ranging from 600 to 1,000 °C and acid-base properties
Lu You et al.
ACS nano, 6(6), 5388-5394 (2012-05-18)
The novel strain-driven morphotropic phase boundary (MPB) in highly strained BiFeO(3) thin films is characterized by well-ordered mixed phase nanodomains (MPNs). Through scanning probe microscopy and synchrotron X-ray diffraction, eight structural variants of the MPNs are identified. Detailed polarization configurations
S Sanghi et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 83(1), 94-99 (2011-09-06)
Glasses with composition 20CdO·xBi(2)O(3)·(79.5-x)B(2)O(3) (15≤x≤35, x in mol%) containing 0.5 mol% of Er(3+) ions were prepared by melt-quench technique (1150°C in air). The amorphous nature of the glasses was confirmed by X-ray diffraction. The spectroscopic properties of the glasses were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service