Skip to Content
Merck
All Photos(3)

Key Documents

205559

Sigma-Aldrich

Ammonium metavanadate

99%

Synonym(s):

Ammonium vanadate(V)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH4VO3
CAS Number:
Molecular Weight:
116.98
EC Number:
MDL number:
UNSPSC Code:
12352300
eCl@ss:
38180409
PubChem Substance ID:
NACRES:
NA.55
grade:
for analytical purposes
form:
powder, crystals or granules

grade

for analytical purposes

Quality Level

Assay

99%

form

powder, crystals or granules

pH

7 (20 °C, 5.1 g/L)

density

2.32 g/cm3 at 25 °C (lit.)

SMILES string

N.O[V](=O)=O

InChI

1S/H3N.H2O.2O.V/h1H3;1H2;;;/q;;;;+1/p-1

InChI key

YBVKNHXQSRDWAA-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Ammonium metavanadate may be employed in the following studies:
  • Preparation of various 2-substituted aryl benzimidazole derivatives.
  • Synthesis of dioxidovanadium(V) complex.
  • Preparation of vanadia modified titania (V2O5/TiO2) photo-catalysts.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Aquatic Chronic 2 - Eye Irrit. 2 - Repr. 2 - STOT RE 1 Inhalation

Target Organs

Respiratory Tract

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Philani Vusumuzi Hlophe et al.
Scientific reports, 9(1), 10044-10044 (2019-07-13)
The development of heterojunctions is the current focus of the scientific community as these materials are visible light active and the staggered positioning of their band edges combats electron-hole recombination which is the downside of most photocatalysts. In this work
Ammonium Metavanadate an Efficient Catalyst for Erlenmeyer Synthesis.
Madje B, et al.
Journal of the Indian Chemical Society, 89(9), 1295-1299 (2012)
Ammonium Metavanadate as an Efficient Catalyst for the Synthesis of 2,4,5-Triaryl-1H-imidazoles.
Niralwad KS, et al.
Journal of Heterocyclic Chemistry, 48(3), 742-745 (2011)
Microwave-assisted one-pot synthesis of octahydroquinazolinone derivatives using ammonium metavanadate under solvent-free condition.
Niralwad KS, et al.
Tetrahedron Letters, 51(28), 3616-3618 (2010)
Ammonium metavanadate: an effective catalyst for synthesis of α-hydroxyphosphonates.
Sonar SS, et al.
ARKIVOC (Gainesville, FL, United States), 2, 138-148 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service