Skip to Content
Merck
All Photos(6)

Key Documents

857890

Sigma-Aldrich

m-Cresol Purple

indicator grade, Dye content 90 %

Synonym(s):

m-Cresolsulfonphthalein

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H18O5S
CAS Number:
Molecular Weight:
382.43
Beilstein:
350314
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

grade

indicator grade

Quality Level

form

powder

composition

Dye content, 90%

technique(s)

titration: suitable

useful pH range

1.2-2.8, red to yellow(acid)
7.4-9.0, yellow to violet(alkaline)

ε (extinction coefficient)

≥ 11000 at 295-301 nm at 0.01 g/L (0.01 G/L in 0.005 N NAOH in Methanol)
≥26000 at 578-584 nm at 0.01 g/L (in 0.005 N NAOH in Methanol)
≥8000 at 369-375 nm at 0.01 g/L (in 0.005 N NAOH in Methanol)

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

Cc1cc(O)ccc1\C(=C2/C=CC(=O)C=C2C)c3ccccc3S(O)(=O)=O

InChI

1S/C21H18O5S/c1-13-11-15(22)7-9-17(13)21(18-10-8-16(23)12-14(18)2)19-5-3-4-6-20(19)27(24,25)26-21/h3-12,22-23H,1-2H3

InChI key

OLQIKGSZDTXODA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

m-Cresol Purple (meta Cresol Purple) is a sulfonephthalein dye. It is also known as m-Cresolsulfonphthalein.

Application

m-Cresol purple has been used to measure pH via spectrophotometer.

Biochem/physiol Actions

m-Cresol purple is a pH indicator. It changes from red (pH 1.2) to yellow (2.8) and yellow (7.4) to purple (9.0).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Relative sensitivity of five Hawaiian coral species to high temperature under high-pCO2 conditions.
Bahr KD, et al.
Coral Reefs, 35, 729-738 (2016)
Seasonal and annual calcification rates of the Hawaiian reef coral, Montipora capitata, under present and future climate change scenarios.
Bahr KD, et al.
ICES Journal of Marine Science, fsw078-fsw078 (2016)
Modifications for SDS-PAGE of proteins.
K G Christy et al.
BioTechniques, 7(7), 692-693 (1989-07-01)
Keisha D Bahr et al.
PloS one, 15(2), e0228168-e0228168 (2020-02-06)
Ocean warming and acidification are among the greatest threats to coral reefs. Massive coral bleaching events are becoming increasingly common and are predicted to be more severe and frequent in the near future, putting corals reefs in danger of ecological
M D Johnston et al.
Journal of applied microbiology, 94(6), 1015-1023 (2003-05-20)
To examine the effect on the leakage of low molecular weight cytoplasmic constituents from Staphylococcus aureus using phenolics singly and in combination, and to see if the observations could be modelled using a non-linear dose response. The rate of potassium

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service