Skip to Content
Merck
All Photos(4)

Key Documents

277908

Sigma-Aldrich

Ammonium molybdate

99.98% trace metals basis

Synonym(s):

Ammonium orthomolybdate, Diammonium molybdate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NH4)2MoO4
CAS Number:
Molecular Weight:
196.01
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
form:
solid
solubility:
ethanol: insoluble(lit.)

Quality Level

Assay

99.98% trace metals basis

form

solid

reaction suitability

reagent type: catalyst
core: molybdenum

technique(s)

electron microscopy: suitable

impurities

≤250.0 ppm Trace Metal Analysis

solubility

ethanol: insoluble(lit.)

SMILES string

N.N.O[Mo](O)(=O)=O

InChI

1S/Mo.2H3N.2H2O.2O/h;2*1H3;2*1H2;;/q+2;;;;;;/p-2

InChI key

LZRXBDCICLHKEN-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Ammonium molybdate can be prepared by dissolving molybdenum trioxide in ammonia.

Application

Ammonium molybdate has been used to prepare phosphomolybdates. (NH4)2MoO4 was used as an analytical reagent for quantitative analysis of phosphates, silicates, arsenates and lead in their aqueous solutions.2 At saturated concentration, (NH4)2MoO4 was used to perform cryo-negative staining and as a negative stain in biological electron microscopy. Ammonium molybdate was used in study of crystallisation of cell-free-expressed membrane proteins.
Ammonium molybdate has been used to prepare phosphomolybdates. (NH4)2MoO4 was used as an analytical reagent for quantitative analysis of phosphates, silicates, arsenates and lead in their aqueous solutions. At saturated concentration, (NH4)2MoO4 was used to perform cryo-negative staining and as a negative stain in biological electron microscopy. Ammonium molybdate was used in study of crystallisation of cell-free-expressed membrane proteins.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Handbook of Non-Ferrous Metal Powders: Technologies and Applications null
Effect of the addition of ammonium molybdate on metakaolin-based geopolymer formation: Shrinkage and crystallization
Vidal L, et al.
Powder Technology, 275, 211-219 (2015)
M Adrian et al.
Micron (Oxford, England : 1993), 29(2-3), 145-160 (1998-07-31)
A procedure is presented for the preparation of thin layers of vitrified biological suspensions in the presence of ammonium molybdate, which we term cryo-negative staining. The direct blotting of sample plus stain solution on holey carbon supports produces thin aqueous
Principles of micro Sequential Injection Analysis in the Lab-on-Valve format and its Introduction into a Teaching Laboratory
Decuir MS, et al.
Journal of Flow Injection Analysis, 24(2), 103-103 (2007)
Leandro Silva Costa et al.
Marine drugs, 9(6), 952-966 (2011-07-13)
Fucan is a term used to denominate a type of polysaccharide which contains substantial percentages of l-fucose and sulfate ester groups. We obtained five heterofucans from Sargassum filipendula by proteolytic digestion followed by sequential acetone precipitation. These heterofucans are composed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service