Skip to Content
Merck
All Photos(1)

Documents

138975

Sigma-Aldrich

1-Pentanol

ReagentPlus®, ≥99%

Synonym(s):

n-Amyl alcohol, Pentyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4OH
CAS Number:
Molecular Weight:
88.15
Beilstein:
1730975
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.21

vapor density

3 (vs air)

Quality Level

vapor pressure

1 mmHg ( 13.6 °C)
1.5 mmHg ( 20 °C)

product line

ReagentPlus®

Assay

≥99%

form

liquid

autoignition temp.

572 °F

expl. lim.

10 %, 100 °F

refractive index

n20/D 1.409 (lit.)

pH

7

bp

136-138 °C (lit.)

mp

−78 °C (lit.)

density

0.811 g/mL at 25 °C (lit.)

SMILES string

CCCCCO

InChI

1S/C5H12O/c1-2-3-4-5-6/h6H,2-5H2,1H3

InChI key

AMQJEAYHLZJPGS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Pentanol is a linear alcohol. Pentane, a ″next generation″ alcohol fuel, is a promising substitute for conventional gasoline and diesel fuels. Kinetics of thermal decomposition of its isomeric forms (1-pentanol, 2-methyl-1-butanol, and 3-methyl-1-butanol) have been reported.
1-Pentanol is a colorless liquid having a pleasant odor, which can be obtained as a product from the process of fractional distillation of mixed alcohols resulting from the chlorination and alkaline hydrolysis of pentane occurs.

Application

1-Pentanol may be used as a cosurfactant in the water-in-oil mixed surfactant microemulsions.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

120.2 °F - closed cup

Flash Point(C)

49 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Harbison.DR, et al.
Hamilton and Hardy's Industrial Toxicology, (6) (2015)
Long Zhao et al.
The journal of physical chemistry. A, 116(37), 9238-9244 (2012-08-23)
Pentanol is one of the promising "next generation" alcohol fuels with high energy density and low hygroscopicity. In the present work, dominant reaction channels of thermal decomposition of three isomers of pentanol: 1-pentanol, 2-methyl-1-butanol, and 3-methyl-1-butanol were investigated by CBS-QB3
Acid/base- and anion-controllable organogels formed from a urea-based molecular switch.
Sheng-Yao Hsueh et al.
Angewandte Chemie (International ed. in English), 49(48), 9170-9173 (2010-10-19)
Experimental and detailed kinetic modeling study of 1-pentanol oxidation in a JSR and combustion in a bomb.
Togbe C, et al.
Proceedings of the Combustion Institute, 33(1), 367-374 (2011)
Ken Nagai et al.
Colloids and surfaces. B, Biointerfaces, 56(1-2), 197-200 (2006-12-16)
We describe here our recent work on spontaneous regular motion of liquid droplet powered by the chemical Marangoni effect under spatially symmetric conditions. It is shown that a spontaneously crawling oil droplet on a glass substrate with a nonequilibrium chemical

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service