Skip to Content
Merck
All Photos(3)

Documents

P8025

Sigma-Aldrich

3-Pentanol

98%

Synonym(s):

sec-inact.-Pentyl alcohol, Diethyl carbinol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)2CHOH
CAS Number:
Molecular Weight:
88.15
Beilstein:
1730964
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

autoignition temp.

815 °F

refractive index

n20/D 1.410 (lit.)

bp

114-115 °C/749 mmHg (lit.)

density

0.815 g/mL at 25 °C (lit.)

SMILES string

CCC(O)CC

InChI

1S/C5H12O/c1-3-5(6)4-2/h5-6H,3-4H2,1-2H3

InChI key

AQIXEPGDORPWBJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3-Pentanol can be used as:
  • A starting material for the preparation of liquid crystals, 1-ethylpropyl (R)-2-[4-(4′-alkoxybiphenylcarbonyloxy)-phenoxy]propionates by reacting with chiral (S)-lactic acid.
  • Solvent/reductant in the catalytic deoxydehydration reaction of C4−C6 sugar alcohols into linear polyene using methyltrioxorhenium as a catalyst.
  • A reactant for the synthesis of 3-(4-bromophenyloxy)pentane by reacting with 4-bromophenol via base-catalyzed Mitsunobu reaction.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

93.2 °F - closed cup

Flash Point(C)

34 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Base catalyzed Mitsunobu reactions as a tool for the synthesis of aryl sec-alkyl ethers
Manivel P, et al.
Tetrahedron Letters, 48(15), 2701-2705 (2007)
Synthesis and mesomorphic properties of chiral liquid crystals derived from (S)-lactic acid with 3-pentanol
Wu S-L and Lin C-Y
Liq. Cryst., 29(12), 1575-1580 (2002)
Deoxygenation of biomass-derived feedstocks: oxorhenium-catalyzed deoxydehydration of sugars and sugar alcohols.
Mika Shiramizu et al.
Angewandte Chemie (International ed. in English), 51(32), 8082-8086 (2012-07-06)
Alfonso Pérez-Garrido et al.
Bioorganic & medicinal chemistry, 17(2), 896-904 (2008-12-06)
This paper reports a QSAR study for predicting the complexation of a large and heterogeneous variety of substances (233 organic compounds) with beta-cyclodextrins (beta-CDs). Several different theoretical molecular descriptors, calculated solely from the molecular structure of the compounds under investigation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service