Skip to Content
Merck
All Photos(1)

Key Documents

51778

Sigma-Aldrich

1-Heptanol

≥99.5% (GC)

Synonym(s):

Heptyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)6OH
CAS Number:
Molecular Weight:
116.20
Beilstein:
1731686
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

0.5 mmHg ( 20 °C)

Assay

≥99.5% (GC)

autoignition temp.

662 °F

refractive index

n20/D 1.424 (lit.)
n20/D 1.424

bp

176 °C (lit.)

mp

−36 °C (lit.)

density

0.822 g/mL at 25 °C (lit.)

functional group

hydroxyl

SMILES string

CCCCCCCO

InChI

1S/C7H16O/c1-2-3-4-5-6-7-8/h8H,2-7H2,1H3

InChI key

BBMCTIGTTCKYKF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

168.8 °F - closed cup

Flash Point(C)

76 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Densities and viscosities for binary mixtures of phenetole with 1-pentanol, 1-hexanol, 1-heptanol, 1-octanol, 1-nonanol, and 1-decanol at different temperatures.
Al-Jimaz AS, et al.
Fluid Phase Equilibria, 218(2), 247-260 (2004)
Volumetric behaviour of acrylic esters (methyl-, ethyl-, and butyl acrylate)+ 1-alcohol (heptanol, octanol, decanol and dodecanol) at 298.15 K and 308.15 K.
Sastry NV and Valand MK.
Physics and Chemistry of Liquids, 38(1), 61-72 (2000)
Dielectric behavior of mixtures of 1-heptanol and 4-heptanol and the fluid structure of the monoalcohols.
Bordewijk P, et al.
The Journal of Physical Chemistry, 73(10), 3255-3258 (1969)
Gabriela Oliveira et al.
International journal of pharmaceutics, 393(1-2), 61-67 (2010-04-08)
While it is well recognised that formulation components influence drug permeation, few studies have addressed the influence of vehicles on drug transport in artificial or biological membranes Previously we have investigated the effects of temperature on the uptake of model
Nannan Ye et al.
Lab on a chip, 11(6), 1096-1101 (2011-02-05)
Transport across gap junction channels (GJCs) between neighboring cells is critical to synchronizing cell's electrical and metabolic activities and maintaining cell homeostasis. Here we present a non-invasive microfluidic method to measure molecular diffusion across GJCs in multiple 1D cell arrays

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service