Skip to Content
Merck
All Photos(2)

Key Documents

574791

Sigma-Aldrich

Hydroxyapatite

synthetic, 99.8% trace metals basis (excludes Mg)

Synonym(s):

Calcium phosphate tribasic, Calcium hydroxyphosphate, HAp, Hydroxylapatite, Tribasic calcium phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[Ca5(OH)(PO4)3]x
CAS Number:
Molecular Weight:
502.31
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99.8% trace metals basis (excludes Mg)

form

solid

impurities

≤2500 ppm Trace Metal Analysis

mp

1100 °C (lit.)

SMILES string

[Ca++].[Ca++].[Ca++].[Ca++].O[Ca+].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O

InChI

1S/5Ca.3H3O4P.H2O/c;;;;;3*1-5(2,3)4;/h;;;;;3*(H3,1,2,3,4);1H2/q5*+2;;;;/p-10

InChI key

XYJRXVWERLGGKC-UHFFFAOYSA-D

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tie Liu et al.
Bone, 56(1), 110-118 (2013-06-05)
As an alternative to an autologous bone graft, deproteinized bovine bone (DBB) is widely used in the clinical dentistry. Although DBB provides an osteoconductive scaffold, it is not capable of enhancing bone regeneration because it is not osteoinductive. In order
Thomas Lyon et al.
The Journal of bone and joint surgery. American volume, 95(23), 2088-2096 (2013-12-07)
Recombinant human bone morphogenetic protein-2 (rhBMP-2) applied on an absorbable collagen sponge improves open tibial fracture-healing as an adjunct to unreamed intramedullary nail fixation. We evaluated rhBMP-2 and a new, injectable calcium phosphate matrix (CPM) formulation in acute closed tibial
Liam M Grover et al.
Biomaterials, 34(28), 6631-6637 (2013-06-12)
Pyrophosphate ions are both inhibitors of HA formation and substrates for phosphatase enzymes. Unlike polyphosphates their hydrolysis results simultaneously in the complete loss of mineral formation inhibition and a localised elevation in orthophosphate ion concentration. Despite recent advances in our
Satyavrata Samavedi et al.
Acta biomaterialia, 9(9), 8037-8045 (2013-06-25)
Calcium phosphate ceramics (CPCs) have been widely used as biomaterials for the regeneration of bone tissue because of their ability to induce osteoblastic differentiation in progenitor cells. Despite the progress made towards fabricating CPCs possessing a range of surface features
W Zhang et al.
Journal of dental research, 92(12), 1136-1141 (2013-10-08)
Stem cell-based bone tissue engineering has been recognized as a new strategy for maxillary sinus floor elevation. More rapid bone formation may enhance this technique when simultaneous dental implant placement is desired. Adipose tissue-derived stem cells (ADSCs) and bone marrow

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service