Skip to Content
Merck
All Photos(1)

Key Documents

02882

Sigma-Aldrich

Ethanol

purum, absolute ethanol, denaturated with 2% 2-butanone, A15 MEK1, ≥99.8% (based on denaturant-free substance)

Synonym(s):

Ethyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2OH
CAS Number:
Molecular Weight:
46.07
Beilstein:
1718733
MDL number:
UNSPSC Code:
15101802
PubChem Substance ID:
NACRES:
NA.05
bp:
78 °C (lit.)
78 °C
vapor pressure:
44.6 mmHg ( 20 °C)
44.6 mmHg
Pricing and availability is not currently available.

vapor density

1.59 (vs air)
1.59 (vs air)

vapor pressure

44.6 mmHg ( 20 °C)
44.6 mmHg

grade

purum

Assay

≥99.8% (based on denaturant-free substance)

form

liquid

autoignition temp.

683 °F

quality

absolute ethanol, denaturated with 2% 2-butanone, A15 MEK1

expl. lim.

19 %, 60 °F
19 %

refractive index

n20/D 1.3600 (lit.)
n20/D 1.363

bp

78 °C (lit.)
78 °C

mp

-114 °C
−114 °C (lit.)

format

neat

SMILES string

CCO
CCO

InChI

1S/C2H6O/c1-2-3/h3H,2H2,1H3

InChI key

LFQSCWFLJHTTHZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Sales restrictions may apply

Footnote

1Distribution and use of ethyl alcohol in Switzerland require a license from the Swiss Federal Alcohol Administration

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

55.4 °F - closed cup

Flash Point(C)

13 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Chandra Kumar Dixit et al.
Nature protocols, 6(4), 439-445 (2011-03-18)
This protocol describes an improved and optimized approach to develop rapid and high-sensitivity ELISAs by covalently immobilizing antibody on chemically modified polymeric surfaces. The method involves initial surface activation with KOH and an O(2) plasma, and then amine functionalization with
Vashist SK.
Protocol Exchange (2011)
A highly-sensitive and rapid Surface Plasmon Resonance immunoassay procedure based on the covalent-orientated immobilization of antibodies.
Vashist SK.
Protocol Exchange (2011)
Spectrophotometric characteristics of chlorophylls a and b and their pheophytins in ethanol.
J F Wintermans et al.
Biochimica et biophysica acta, 109(2), 448-453 (1965-11-29)
A new oxidase-peroxidase kit for ethanol assays in alcoholic beverages.
Gonchar MV, et al.
Food Technology and Biotechnology, 39(1), 37-42 (2001)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service