Skip to Content
Merck
All Photos(1)

Key Documents

Y0001306

Cefpodoxime proxetil for peak identification

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Cefpodoxime proxetil

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H27N5O9S2
CAS Number:
Molecular Weight:
557.60
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

cefpodoxime

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

SMILES string

[s]1c(nc(c1)\C(=N\OC)\C(=O)N[C@H]2[C@H]3SCC(=C(N3C2=O)C(=O)OC(OC(=O)OC(C)C)C)COC)N

InChI

1S/C21H27N5O9S2/c1-9(2)33-21(30)35-10(3)34-19(29)15-11(6-31-4)7-36-18-14(17(28)26(15)18)24-16(27)13(25-32-5)12-8-37-20(22)23-12/h8-10,14,18H,6-7H2,1-5H3,(H2,22,23)(H,24,27)/b25-13-/t10?,14-,18-/m1/s1

InChI key

LTINZAODLRIQIX-FBXRGJNPSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Cefpodoxime proxetil for peak identification EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vasu Kumar Kakumanu et al.
Yakugaku zasshi : Journal of the Pharmaceutical Society of Japan, 128(3), 439-445 (2008-03-04)
Cefpodoxime proxetil (CP) is a prodrug with poor oral bioavailability because of its metabolism to Cefpodoxime acid (CA) in luminal contents and intestinal epithelial cells. In the present investigation, regional variability in different segments of the gastrointestinal tract vis-à-vis solubility
Junho Chu et al.
International journal of pharmaceutics, 369(1-2), 85-91 (2008-12-02)
Fine particles of cefpodoxime proxetil (CPD) were prepared using an Aerosol Solvent Extraction System (ASES) with supercritical CO(2). The resulting primary particles were approximately 0.1-0.2microm in size and were almost spherical in shape. The secondary particles were approximately 0.2-0.6microm in
Naoto Fukutsu et al.
Journal of chromatography. A, 1129(2), 153-159 (2006-07-19)
Application of the HPLC hyphenated techniques of LC-MS, LC-NMR and solvent-elimination LC-IR was demonstrated by the identification of the degradation products of a third generation cephalosporin antibiotic, cefpodoxime proxetil, in solid state, drug formulation and solution. Molecular weight and fragment
Amrita Bajaj et al.
Drug development and industrial pharmacy, 39(5), 635-645 (2012-05-09)
Lipid based drug delivery systems have gained prominence in last decade for drugs with dissolution rate limited oral bioavailability. To improve the solubility, permeability and oral bioavailability of cefpodoxime proxetil, β-lactam antibiotic. It is BCS Class IV drug having solubility
Md Salim Shakur et al.
Indian pediatrics, 44(11), 838-841 (2007-12-07)
In order to evaluate clinical and bacteriological efficacy of Cefpodoxime Proxetil (CP) in typhoid fever in comparison to cefixime (CF), we assessed 140 children with suspected typhoid fever. Fulfilling inclusion criteria finally 40 culture confirmed typhoid fever were allocated in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service