Skip to Content
Merck
All Photos(1)

Documents

46738

Supelco

Spectinomycin dihydrochloride pentahydrate

VETRANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H24N2O7 · 2HCl · 5H2O
CAS Number:
Molecular Weight:
495.35
Beilstein:
5684150
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

solubility

H2O: soluble at 

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria

application(s)

clinical testing

format

neat

Mode of action

protein synthesis | interferes

storage temp.

2-8°C

SMILES string

Cl[H].Cl[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H][C@@]12O[C@H](C)CC(=O)[C@]1(O)O[C@]3([H])[C@@H](NC)[C@@H](O)[C@@H](NC)[C@H](O)[C@@]3([H])O2

InChI

1S/C14H24N2O7.2ClH.5H2O/c1-5-4-6(17)14(20)13(21-5)22-12-10(19)7(15-2)9(18)8(16-3)11(12)23-14;;;;;;;/h5,7-13,15-16,18-20H,4H2,1-3H3;2*1H;5*1H2/t5-,7-,8+,9+,10+,11-,12-,13+,14+;;;;;;;/m1......./s1

InChI key

DCHJOVNPPSBWHK-UXXUFHFZSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: aminoglycoside

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Spectinomycin dihydrochloride pentahydrate may be used as a reference standard for the determination of spectinomycin in bovine milk samples by liquid chromatography with electrochemical detection.
Spectinomycin is an aminocyclitol antibiotic derived from Streptomyces spectabilis. It has been shown to have a wide variety of uses, including treating acute gonorrheal urethritis and cervictis, to mark cell layers to monitor cell fate during leaf development, as a selection marker in plant related transformation systems for plant cells containing the marker gene Spcr, to study respiratory tract infections of cattle caused by Pasteurella multocida and Mannheimia haemolytica, and to generate plants deficient for the plastid-encoded RNA polymerase on MS-agar media. Spectinomycin also has use in amplification of low copy number plasmid carrying replicons. It is recommended for use in cell culture applications at 7.5-20 mg/L.

Caution

Solutions can be stored at 2-8°C for several weeks or at -20°C for more extended periods.

Preparation Note

Stock solutions should be prepared at 10 mg/mL in water and filter sterilized.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of spectinomycin residues in bovine milk using liquid chromatography with electrochemical detection.
Schermerhorn PG, et al.
Journal of Agricultural and Food Chemistry, 43(8), 2122- 2125 (1995)
Liu-Feng Yuan et al.
Sexually transmitted diseases, 38(8), 764-768 (2011-08-17)
Antimicrobial resistance in Neisseria gonorrhoeae (NG) is a public health concern globally, and azithromycin-resistant NG isolates have been identified in many countries. This study aimed to investigate azithromycin susceptibility in NG and to genetically characterize the azithromycin-resistant isolates identified in
E A Filipenko et al.
Genetika, 47(1), 41-47 (2011-03-31)
Bioballistic transformation of carrot Daucus carota L. callus cultures with a plasmid containing the aadA (aminoglycoside 3'-adenyltransferase) gene and subsequent selection oftransformants on a selective medium containing spectinomycin (100-500 mg/l) yielded ten callus lines resistant to this antibiotic. PCR analysis
Tarinee Tungsuchat-Huang et al.
Plant molecular biology, 76(3-5), 453-461 (2011-01-05)
Identification of a genetically stable Nicotiana tabacum (tobacco) plant with a uniform population of transformed plastid genomes (ptDNA) takes two cycles of plant regeneration from chimeric leaves and analysis of multiple shoots by Southern probing in each cycle. Visual detection
Natividad Cabrera-Valladares et al.
Journal of molecular microbiology and biotechnology, 22(3), 177-197 (2012-08-01)
The phosphoenolpyruvate (PEP) node is an important carbon distribution point in the central metabolic networks; therefore, its modification is a common strategy employed for developing microbial production strains. In this study, mutants of Bacillus subtilis 168 were generated with deletions

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service