Skip to Content
Merck
All Photos(1)

Key Documents

33429

Supelco

Doxycycline hyclate

VETRANAL®, analytical standard

Synonym(s):

Doxycycline hydrochloride hemiethanolate hemihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H24N2O8 · HCl · 0.5H2O · 0.5C2H6O
CAS Number:
Molecular Weight:
512.94
Beilstein:
5702728
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

Agency

EPA 1694

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

solubility

DMSO: soluble 100 mM

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria
mycoplasma
parasites

application(s)

clinical testing

format

neat

Mode of action

protein synthesis | interferes

SMILES string

Cl[H].Cl[H].[H]O[H].CCO.C[C@@H]1C2[C@H](O)C3[C@H](N(C)C)C(O)=C(C(N)=O)C(=O)[C@@]3(O)C(O)=C2C(=O)c4c(O)cccc14.C[C@@H]5C6[C@H](O)C7[C@H](N(C)C)C(O)=C(C(N)=O)C(=O)[C@@]7(O)C(O)=C6C(=O)c8c(O)cccc58

InChI

1S/2C22H24N2O8.C2H6O.2ClH.H2O/c2*1-7-8-5-4-6-9(25)11(8)16(26)12-10(7)17(27)14-15(24(2)3)18(28)13(21(23)31)20(30)22(14,32)19(12)29;1-2-3;;;/h2*4-7,10,14-15,17,25,27-29,32H,1-3H3,(H2,23,31);3H,2H2,1H3;2*1H;1H2/t2*7-,10?,14?,15-,17-,22-;;;;/m00..../s1

InChI key

HALQELOKLVRWRI-ZVACAFRPSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: tetracycline

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Mode of Action: Doxycycline hyclate inhibits the inflammatory response to the Lyme Disease Spirochete Borrelia burgdorferi.

Activity Spectrum: Effective against a broad spectrum inhibitor of matrix metalloproteinases in vivo.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Erdinc Turk et al.
Animals : an open access journal from MDPI, 10(6) (2020-07-01)
The aims of this study in goats were to determine the pharmacokinetics of doxycycline hyclate following single intravenous (IV), intramuscular (IM) and oral administrations of 20 mg/kg and to evaluate the pharmacokinetics and accumulation of doxycycline hyclate after repeated oral
Hanumara Ram Chowdri et al.
Annals of internal medicine, 159(1), 21-27 (2013-07-03)
The diverse tickborne infections of the northeastern United States can present as undifferentiated flu-like illnesses. In areas endemic for Lyme and other tickborne diseases, patients presenting with acute febrile illness with myalgia, headache, neutropenia, thrombocytopenia, and elevated hepatic aminotransferase levels
C Arnoud Meijer et al.
Annals of internal medicine, 159(12), 815-823 (2014-02-04)
Doxycycline inhibits formation and progression of abdominal aortic aneurysms (AAAs) in preclinical models of the disease, but it is unclear whether and how this observation translates to humans. To test whether doxycycline inhibits AAA progression in humans. Randomized, placebo-controlled, double-blind
Xuefeng Wu et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(38), 13870-13875 (2014-09-06)
Metastatic spread is the leading cause of cancer mortality. Breast cancer (BCa) metastatic recurrence can happen years after removal of the primary tumor. Here we show that Ubc13, an E2 enzyme that catalyzes K63-linked protein polyubiquitination, is largely dispensable for
Alicia Anderson et al.
MMWR. Recommendations and reports : Morbidity and mortality weekly report. Recommendations and reports, 62(RR-03), 1-30 (2013-03-29)
Q fever, a zoonotic disease caused by the bacterium Coxiella burnetii, can cause acute or chronic illness in humans. Transmission occurs primarily through inhalation of aerosols from contaminated soil or animal waste. No licensed vaccine is available in the United

Questions

1–2 of 2 Questions  
  1. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  2. Is doxycycline hyclate soluble in DMSO?

    1 answer
    1. According to literature sources, doxycycline hyclate is soluble in DMSO at a concentration of 100 mM.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service