Skip to Content
Merck
All Photos(1)

Documents

Y0001111

Ceftazidime for peak identification

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Ceftazidime pentahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H22N6O7S2 · 5H2O
CAS Number:
Molecular Weight:
636.65
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

ceftazidime

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

InChI

1S/C22H22N6O7S2.5H2O/c1-22(2,20(33)34)35-26-13(12-10-37-21(23)24-12)16(29)25-14-17(30)28-15(19(31)32)11(9-36-18(14)28)8-27-6-4-3-5-7-27;;;;;/h3-7,10,14,18H,8-9H2,1-2H3,(H4-,23,24,25,29,31,32,33,34);5*1H2/b26-13-;;;;;/t14-,18-;;;;;/m1...../s1

InChI key

NMVPEQXCMGEDNH-TZVUEUGBSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Ceftazidime for peak identification EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Covalent trapping and bacterial resistance to ceftazidime.
Jean-Marie A Frère
The Journal of biological chemistry, 288(37), 26967-26967 (2013-09-17)
María M Tavío et al.
Journal of medical microbiology, 63(Pt 1), 56-65 (2013-10-04)
The mechanisms responsible for the increase in ceftazidime MIC in two Escherichia coli in vitro selected mutants, Caz/20-1 and Caz/20-2, were studied. OmpF loss and overexpression of acrB, acrD and acrF that were associated with acrR and marR mutations and
Jingshu Ji et al.
Antimicrobial agents and chemotherapy, 57(11), 5697-5700 (2013-08-14)
It is unclear whether the genetic background of drug-resistant Pseudomonas aeruginosa was disseminated from a certain clone. Thus, we performed MLST (multilocus sequence typing) of 896 P. aeruginosa isolates that were nonsusceptible to imipenem, meropenem, or ceftazidime. This revealed 254
Ayman M Goudah et al.
Journal of the American Veterinary Medical Association, 243(3), 424-429 (2013-07-20)
To determine the plasma disposition kinetics, absolute bioavailability, and milk concentrations of ceftazidime in healthy lactating female dromedary camels (Camelus dromedarius) following IV and IM administration of a single dose of 10 mg/kg (4.5 mg/lb). Prospective crossover study. 8 healthy
First outbreak of KPC-3-producing Klebsiella pneumoniae (ST258) clinical isolates in a Mexican Medical Center.
Patricia Rodríguez-Zulueta et al.
Antimicrobial agents and chemotherapy, 57(8), 4086-4088 (2013-06-05)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service