Skip to Content
Merck
All Photos(1)

Key Documents

C0690500

Ceftazidime

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Ceftazidime pentahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H22N6O7S2 · 5H2O
CAS Number:
Molecular Weight:
636.65
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

ceftazidime

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[s]1c(nc(c1)\C(=N\OC(C)(C)C(=O)[O-])\C(=O)N[C@H]2[C@H]3SCC(=C(N3C2=O)C(=O)O)C[n+]4ccccc4)N.O.O.O.O.O

InChI

1S/C22H22N6O7S2.5H2O/c1-22(2,20(33)34)35-26-13(12-10-37-21(23)24-12)16(29)25-14-17(30)28-15(19(31)32)11(9-36-18(14)28)8-27-6-4-3-5-7-27;;;;;/h3-7,10,14,18H,8-9H2,1-2H3,(H4-,23,24,25,29,31,32,33,34);5*1H2/b26-13-;;;;;/t14-,18-;;;;;/m1...../s1

InChI key

NMVPEQXCMGEDNH-TZVUEUGBSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Ceftazidime EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anu Kantele et al.
Clinical infectious diseases : an official publication of the Infectious Diseases Society of America, 60(6), 837-846 (2015-01-24)
More than 300 million travelers visit regions with poor hygiene annually. A significant percentage of them become colonized by resistant intestinal bacteria such as extended-spectrum beta-lactamase-producing Enterobacteriaceae (ESBL-PE) and may transmit the strains to others and to medical care settings
Johanna Berkhout et al.
Antimicrobial agents and chemotherapy, 59(2), 1138-1144 (2014-12-10)
To evaluate the in vitro effects of the combination of ceftazidime and avibactam on the MICs of both compounds, checkerboard assays were performed for 81 clinical strains, including 55 Enterobacteriaceae strains (32 Klebsiella pneumoniae, 19 Escherichia coli, 1 Citrobacter freundii
George G Zhanel et al.
Drugs, 73(2), 159-177 (2013-02-02)
Avibactam (formerly NXL104, AVE1330A) is a synthetic non-β-lactam, β-lactamase inhibitor that inhibits the activities of Ambler class A and C β-lactamases and some Ambler class D enzymes. This review summarizes the existing data published for ceftazidime-avibactam, including relevant chemistry, mechanisms
A Walkty et al.
Antimicrobial agents and chemotherapy, 57(11), 5707-5709 (2013-08-14)
The in vitro activity of ceftolozane in combination with tazobactam (fixed concentration of 4 μg/ml) was evaluated against 2,435 Pseudomonas aeruginosa clinical isolates obtained from across Canada using Clinical and Laboratory Standards Institute broth microdilution methods. The MIC50 and MIC90
Gavin C K W Koh et al.
PLoS neglected tropical diseases, 7(10), e2500-e2500 (2013-10-23)
Burkholderia pseudomallei infection (melioidosis) is an important cause of community-acquired Gram-negative sepsis in Northeast Thailand, where it is associated with a ~40% mortality rate despite antimicrobial chemotherapy. We showed in a previous cohort study that patients taking glyburide ( =

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service