Skip to Content
Merck
All Photos(1)

Documents

C2100000

Cholecalciferol

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

(+)-Vitamin D3, 7-Dehydrocholesterol activated, Activated 7-dehydrocholesterol, Calciol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H44O
CAS Number:
Molecular Weight:
384.64
Beilstein:
2339331
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

biological source

synthetic

grade

pharmaceutical primary standard

Agency

EP

API family

cholecalciferol

form

solid

manufacturer/tradename

EDQM

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

mp

83-86 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

SMILES string

CC(C)CCC[C@@H](C)[C@@]1([H])CC[C@@]([C@]1(C)CCC/2)([H])C2=C\C=C(C[C@@H](O)CC3)/C3=C

InChI

1S/C27H44O/c1-19(2)8-6-9-21(4)25-15-16-26-22(10-7-17-27(25,26)5)12-13-23-18-24(28)14-11-20(23)3/h12-13,19,21,24-26,28H,3,6-11,14-18H2,1-2,4-5H3/b22-12+,23-13-/t21-,24+,25-,26+,27-/m1/s1

InChI key

QYSXJUFSXHHAJI-YRZJJWOYSA-N

Gene Information

human ... VDR(7421)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Cholecalciferol is a fat soluble vitamin, found commercially in vitamin supplements and rodenticides. It can be directly produced on exposure to sunlight or through normal dietary intake.

Application

This European Pharmacopoeia reference standard is intended for use only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Vitamin D acts through a receptor that is a member of the ligand-dependent transcription factor superfamily. Modulates the proliferation and differentiation of both normal and cancer cells. Has antiproliferative and antimetastatic effects on breast, colon, and prostate cancer cells. Activated vitamin D receptors in intestine and bone maintain calcium absorbance and homeostasis.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - STOT RE 1 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Clinical Veterinary Toxicology (2003)
Simultaneous determination of retinol acetate, retinol palmitate, cholecalciferol, α-tocopherol acetate and alphacalcidol in capsules by non-aqueous reversed-phase HPLC and column backflushing
Wielinski S and Olszanowski A
Chromatographia, 50(1-2), 109-112 (1999)
Sara S Oberhelman et al.
Mayo Clinic proceedings, 88(12), 1378-1387 (2013-12-03)
To determine whether a single monthly supplement is as effective as a daily maternal supplement in increasing breast milk vitamin D to achieve vitamin D sufficiency in their infants. Forty mothers with exclusively breast-fed infants were randomized to receive oral
Bich Tran et al.
The American journal of clinical nutrition, 99(1), 156-161 (2013-10-11)
Observational data suggested that supplementation with vitamin D could reduce risk of infection, but trial data are inconsistent. We aimed to examine the effect of oral vitamin D supplementation on antibiotic use. We conducted a post hoc analysis of data
Ian R Reid et al.
Lancet (London, England), 383(9912), 146-155 (2013-10-15)
Findings from recent meta-analyses of vitamin D supplementation without co-administration of calcium have not shown fracture prevention, possibly because of insufficient power or inappropriate doses, or because the intervention was not targeted to deficient populations. Despite these data, almost half

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service