Skip to Content
Merck
All Photos(1)

Key Documents

Y0001053

Crospovidone

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Polyvinylpyrrolidone, PVP, Polyvidone, Povidone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H9NO)n
CAS Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

crospovidone

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

C=CN1CCCC1=O

InChI

1S/C6H9NO/c1-2-7-5-3-4-6(7)8/h2H,1,3-5H2

InChI key

WHNWPMSKXPGLAX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Crospovidone EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Polyvinylpyrrolidone is a component of Denhardt′s Solution and is included at a concentration of 1% (w/v) in the standard 50X stock solution.
Sales restrictions may apply.

related product

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Polyvinylpyrrolidone (PVP), its diagnostic, therapeutic and technical application and consequences thereof.
W Wessel et al.
Arzneimittel-Forschung, 21(10), 1468-1482 (1971-10-01)
Atsuko Adachi et al.
Contact dermatitis, 48(3), 133-136 (2003-05-21)
A 59-year-old woman who had had several episodes of contact urticaria after hair treatment, developed anaphylaxis after vaginal application of povidone-iodine solution for disinfection. Prick tests showed wheal-and-flare responses to both povidone-iodine (0.1% aqueous) and polyvinylpyrrolidone (povidone, PVP) (0.001% aq.)
Patricia C Buchsel
Expert opinion on drug metabolism & toxicology, 4(11), 1449-1454 (2008-10-28)
Painful oral lesions in patients with cancer, HIV or aphthous ulcers can cause significant pain and discomfort. Those with oral mucositis (OM) resulting from high-dose chemotherapy or irradiation are the most at risk for painful lesions that lead to significant
[Gordon's polyvinylpyrrolidone test].
J Aebersold
Helvetica medica acta, 32(3), 227-246 (1965-09-01)
Povidone-iodine solutions in traumatic wound preparation.
M S Oberg
The American journal of emergency medicine, 5(6), 553-555 (1987-11-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service