Skip to Content
Merck
All Photos(1)

Key Documents

Y0000714

Ritonavir

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

1,3-Thiazol-5-ylmethyl N-[(2S,3S,5S)-3-hydroxy-5-[[(2S)-3-methyl-2-[[methyl-[(2-propan-2-yl-1,3-thiazol-4-yl)methyl]carbamoyl]amino]butanoyl]amino]-1,6-diphenylhexan-2-yl]carbamate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C37H48N6O5S2
CAS Number:
Molecular Weight:
720.94
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

biological source

synthetic

grade

pharmaceutical primary standard

Agency

EP

API family

ritonavir

packaging

pkg of 80 mg

manufacturer/tradename

EDQM

storage condition

protect from light

solubility

ethanol: soluble
methanol: soluble

application(s)

pharmaceutical (small molecule)

format

neat

shipped in

ambient

storage temp.

2-8°C

SMILES string

[s]1c(nc(c1)CN(C)C(=O)N[C@@H](C(C)C)C(=O)N[C@H](C[C@H](O)[C@@H](NC(=O)OCc4[s]cnc4)Cc3ccccc3)Cc2ccccc2)C(C)C

InChI

1S/C37H48N6O5S2/c1-24(2)33(42-36(46)43(5)20-29-22-49-35(40-29)25(3)4)34(45)39-28(16-26-12-8-6-9-13-26)18-32(44)31(17-27-14-10-7-11-15-27)41-37(47)48-21-30-19-38-23-50-30/h6-15,19,22-25,28,31-33,44H,16-18,20-21H2,1-5H3,(H,39,45)(H,41,47)(H,42,46)/t28-,31-,32-,33-/m0/s1

InChI key

NCDNCNXCDXHOMX-XGKFQTDJSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Ritonavir belongs to the group of protease inhibitors that are widely used in combination with other drugs in the prevention of HIV. Its mode of action involves binding to the active site of the protease enzyme and preventing the further maturation of new viral particles.[1]

Application

This European Pharmacopoeia reference standard is intended for use only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mónica M Calderón et al.
Pharmacotherapy, 34(11), 1151-1158 (2014-08-22)
Panax ginseng has been shown in preclinical studies to modulate cytochrome P450 enzymes involved in the metabolism of HIV protease inhibitors. Therefore, the purpose of this study was to determine the influence of P. ginseng on the pharmacokinetics of the
Alicia Khululwa Khwitshana et al.
South African medical journal = Suid-Afrikaanse tydskrif vir geneeskunde, 104(7), 495-500 (2014-09-13)
HIV research is a therapeutic area for which well-defined population-specific treatment and prophylaxis guidelines exist. However, there are limited objective, evidence-based data for assessing adherence to these guidelines. To conduct a retrospective, cross sectional study of adult HIV-infected patients receiving
RP-HPLC method development and validation for simultaneous estimation of Lopinavir and Ritonavir in tablet dosage form
Bindu MJ, et al.
International Journal of Science and Engineering Applications, 5(1), 77-77 (2017)
HPTLC method for simultaneous determination of lopinavir and ritonavir in capsule dosage form
Sulebhavikar VA, et al.
Journal of Chemistry, 5(4), 706-712 (2008)
Venkateswaran C Pillai et al.
Journal of clinical pharmacology, 54(11), 1272-1279 (2014-05-23)
Nilotinib is used to treat chronic myeloid leukemia (CML), and is metabolized by CYP3A. With a black-box warning for QT prolongation, which is exposure dependent, controlling for drug interactions is clinically relevant. Treatments of HIV patients with CML are with

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service