Skip to Content
Merck
All Photos(1)

Key Documents

Y0000639

Cladribine

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

2-Chloro-2′-deoxyadenosine, CdA, Cladribine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12ClN5O3
CAS Number:
Molecular Weight:
285.69
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

cladribine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

SMILES string

Nc1nc(Cl)nc2n(cnc12)[C@H]3C[C@H](O)[C@@H](CO)O3

InChI

1S/C10H12ClN5O3/c11-10-14-8(12)7-9(15-10)16(3-13-7)6-1-4(18)5(2-17)19-6/h3-6,17-18H,1-2H2,(H2,12,14,15)/t4-,5+,6+/m0/s1

InChI key

PTOAARAWEBMLNO-KVQBGUIXSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Cladribine EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Muta. 2 - Repr. 2 - STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thomas P Leist et al.
Clinical neuropharmacology, 34(1), 28-35 (2011-01-19)
Multiple sclerosis (MS) is an inflammatory neurodegenerative disease of the central nervous system. The inflammation is driven significantly by autoreactive lymphocytes, which recruit cells of the innate immune system such as macrophages that contribute to subsequent tissue damage, ultimately resulting
Hans-Peter Hartung et al.
Journal of neurology, 257(2), 163-170 (2009-11-19)
Multiple sclerosis (MS) is a chronic immune-mediated disorder of the CNS in which autoreactive CD4+ and CD8+ T lymphocytes, B lymphocytes, antibodies, macrophages and cytokines synergize to attack myelin sheaths and injure underlying axons. Current disease-modifying drugs (DMDs) for MS
Melissa L Torrey et al.
Leukemia & lymphoma, 52 Suppl 2, 29-33 (2011-04-21)
Hairy cell leukemia (HCL) is an indolent, mature B-cell lymphoproliferative malignancy that clinically manifests as pancytopenia and fibrosis of the bone marrow, ultimately leading to recurrent infections and splenomegaly. This article reviews the extensive experience at Scripps Clinic with cladribine
Martin S Tallman
Leukemia & lymphoma, 52 Suppl 2, 65-68 (2011-04-06)
Hairy cell leukemia (HCL) is a highly treatable, but generally incurable B-cell lymphoproliferative disorder with a long natural history. The purine analogs cladribine and pentostatin are the treatments of choice and both induce complete remission (CR) by peripheral blood counts
James B Johnston
Leukemia & lymphoma, 52 Suppl 2, 43-45 (2011-04-06)
Pentostatin (2'-deoxycoformycin; dCF) and cladribine (2-chlorodeoxyadenosine; CdA) are highly effective agents for the treatment of hairy cell leukemia. Although their precise mechanisms of action in this disease are still unknown, a number of mechanisms have been postulated. dCF is a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service