Skip to Content
Merck

Skip To

F2773

2-Fluoroadenine-9-β-D-arabinofuranoside

DNA synthesis inhibitor

Synonym(s):

9-β-D-Arabinofuranosyl-2-fluoroadenine, F-ara-A, Fludarabine des-phosphate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C10H12FN5O4
CAS Number:
Molecular Weight:
285.23
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352202
EC Number:
244-525-5
MDL number:
Beilstein/REAXYS Number:
1225932
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

2-Fluoroadenine-9-β-D-arabinofuranoside, DNA synthesis and methylation inhibitor

Quality Segment

storage temp.

2-8°C

SMILES string

Nc1nc(F)nc2n(cnc12)[C@@H]3O[C@H](CO)[C@@H](O)[C@@H]3O

InChI

1S/C10H12FN5O4/c11-10-14-7(12)4-8(15-10)16(2-13-4)9-6(19)5(18)3(1-17)20-9/h2-3,5-6,9,17-19H,1H2,(H2,12,14,15)/t3-,5-,6+,9-/m1/s1

InChI key

HBUBKKRHXORPQB-FJFJXFQQSA-N

Gene Information

Application

2-Fluoroadenine-9-β-D-arabinofuranoside (F-ara-A) has been used:
  • to assess its interaction with kinase inhibitor UCN-01 in human leukemia cells (U937 and HL-60) and primary patient samples
  • to assess its influence on endothelial cells by apoptosis assay and human microvascular endothelial cells (HMECs) by cytotoxicity assay
  • in comparative cytotoxicity studies with Alemtuzumab CDC in 17p13- chronic lymphocytic leukemia (CLL) patients

Biochem/physiol Actions

Fludarabine (the 5′-phosphate) is a prodrug that is converted to F-ara-A, which enters cells and accumulates primarily as the 5′-triphosphate. F-ara-A interferes with DNA synthesis and repair and induces apoptosis of cancer cells.
Fludarabine (the 5′-phosphate) is a prodrug that is converted to F-ara-A, which enters cells and accumulates primarily as the 5′-triphosphate. F-ara-A interferes with DNA synthesis and repair and induces apoptosis of cancer cells. F-ara-A also strongly inhibits DNA methylation, particularly methylation of cytosine in CpG dinucleotide sequences.

Disclaimer

The name fludarabine refers to 9-β-D-arabinofuranosyl-2-fluoroadenine 5′-phosphate, but is sometimes erroneously used for this compound, which lacks the phosphate.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
F9813Y00022561272204
Quality Level

200

Quality Level

200

Quality Level

-

Quality Level

-

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

2-8°C

Gene Information

human ... ADORA3(140)
rat ... Adora1(29290), Adora2a(25369), Adora3(25370)

Gene Information

-

Gene Information

-

Gene Information

-


pictograms

Health hazard

signalword

Warning

Hazard Classifications

Muta. 2 - Repr. 2

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Candida Vitale et al.
Scientific reports, 13(1), 16950-16950 (2023-10-08)
Despite recent relevant therapeutic progresses, chronic lymphocytic leukemia (CLL) remains an incurable disease. Selinexor, an oral inhibitor of the nuclear export protein XPO1, is active as single agent in different hematologic malignancies, including CLL. The purpose of this study was
Interactions between 2-fluoroadenine 9-beta-d-arabinofuranoside and the kinase inhibitor UCN-01 in human leukemia and lymphoma cells
Harvey S, et al.
Clinical Cancer Research, 7(2), 320-330 (2001)
Fludarabine induces apoptosis, activation, and allogenicity in human endothelial and epithelial cells: protective effect of defibrotide
Eissner G, et al.
Blood, 100(1), 334-340 (2002)



Global Trade Item Number

SKUGTIN
F2773-5MG04061832947549
F2773-10MG04061832947532

Questions

Reviews

No rating value

Active Filters