Skip to Content
Merck
All Photos(1)

Key Documents

Y0000609

Cladribine for peak identification

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

2-Chloro-2′-deoxyadenosine, CdA, Cladribine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12ClN5O3
CAS Number:
Molecular Weight:
285.69
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

cladribine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

Nc1nc(Cl)nc2n(cnc12)[C@H]3C[C@H](O)[C@@H](CO)O3

InChI

1S/C10H12ClN5O3/c11-10-14-8(12)7-9(15-10)16(3-13-7)6-1-4(18)5(2-17)19-6/h3-6,17-18H,1-2H2,(H2,12,14,15)/t4-,5+,6+/m0/s1

InChI key

PTOAARAWEBMLNO-KVQBGUIXSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Cladribine for peak identification EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Muta. 2 - Repr. 2 - STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

V Singh et al.
Clinical and experimental immunology, 173(2), 288-297 (2013-04-24)
2-Chlorodeoxyadenosine (cladribine, CdA) is an immunosuppressive drug that is licensed to treat hairy cell leukaemia, and has been shown recently to have beneficial effects in patients with multiple sclerosis (MS). The therapeutic effects of CdA have been suggested to be
Joshua D Rosenberg et al.
Blood, 123(2), 177-183 (2013-11-07)
Hairy cell leukemia (HCL) is a rare, indolent B-cell disorder in which single courses of cladribine induce high rates of complete responses. We report on 88 young HCL patients (≤40 years of age at diagnosis) treated with cladribine from the
Philippa L Kohnke et al.
Journal of proteome research, 11(9), 4436-4448 (2012-07-31)
Fludarabine and cladribine are purine analogues used to treat hematological malignancies. Alone or in combination with therapeutic antibodies, they are effective in treating patients with chronic lymphocytic leukemia and non-Hodgkin's lymphoma. However, the mechanisms of action of these drugs are
Tomasz Bączek et al.
Journal of pharmaceutical and biomedical analysis, 70, 330-336 (2012-06-05)
Individualization of the topotecan dosing can reduce inter-patient variability, toxicity, and at the same time increases chemotherapy efficacy. Topotecan dosing based on simultaneous drug monitoring and pharmacokinetic analysis can yield more accurate and precise estimation of the topotecan systemic exposure
Darren S Sigal et al.
Blood, 116(16), 2884-2896 (2010-07-17)
Before the contemporary development of rationally designed antineoplastic therapies, cladribine was identified as a lymphocyte-specific agent. Its profound impact on the natural history of hairy cell leukemia, with responses approaching 100% and a median duration of response of nearly a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service