Skip to Content
Merck
All Photos(1)

Key Documents

S0700000

Sodium calcium edetate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Ethylenediaminetetraacetic acid calcium disodium salt, (Ethylenedinitrilo)tetraacetic acid calcium disodium salt, EDTA calcium disodium salt, Edathamil

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H12N2O8CaNa2
CAS Number:
Molecular Weight:
374.27
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

edta

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[Na+].[Na+].[Ca++].[O-]C(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O

InChI

1S/C10H16N2O8.Ca.2Na/c13-7(14)3-11(4-8(15)16)1-2-12(5-9(17)18)6-10(19)20;;;/h1-6H2,(H,13,14)(H,15,16)(H,17,18)(H,19,20);;;/q;+2;2*+1/p-4

InChI key

SHWNNYZBHZIQQV-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Used to eliminate enzyme inhibition by traces of heavy metals, and to inhibit enzymes that require divalent cations as cofactors. As this form is already chelated with Ca2+, it is selective for divalent cations with greater affinity for EDTA, which it will exchange for Ca2+.
Used to eliminate inhibition of enzyme catalyzed reactions due to traces of heavy metals

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sally Bradberry et al.
Clinical toxicology (Philadelphia, Pa.), 47(9), 841-858 (2009-10-27)
This article reviews the experimental and clinical studies that have compared the efficacy (impact on urine lead excretion, blood and tissue lead concentrations, resolution of features and survival) of sodium calcium edetate (edetate calcium disodium) and succimer (DMSA) in the
David Voglar et al.
Environmental technology, 35(9-12), 1389-1400 (2014-04-08)
We demonstrate here, in a pilot-scale experiment, the feasibility of ethylenediaminetetraacetate (EDTA)based washing technology for soils contaminated with potentially toxic metals. Acid precipitation coupled to initial alkaline toxic metal removal and an electrochemical advanced oxidation process were used for average
Il Joong Park et al.
Archives of pathology & laboratory medicine, 138(8), 1077-1082 (2014-07-31)
The Samsung LABGEO(HC10) Hematology Analyzer (LABGEO(HC10)) is a recently developed automated hematology analyzer that uses impedance technologies. The analyzer provides 18 parameters including 3-part differential at a maximum rate of 80 samples per hour. To evaluate the performance of the
Trial to Assess Chelation Therapy (TACT) and equipoise: When evidence conflicts with beliefs.
David J Maron et al.
American heart journal, 168(1), 4-5 (2014-06-24)
Pseudothrombocytopenia in a patient receiving romiplostim for immune thrombocytopenic purpura.
Claudio Fozza et al.
Annals of hematology, 93(5), 899-900 (2013-10-02)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service