Skip to Content
Merck
All Photos(1)

Key Documents

T2550000

Tris Base

solid, pharmaceutical primary standard, European Pharmacopoeia (EP) Reference standard

Synonym(s):

Tromethamine, 2-Amino-2-(hydroxymethyl)-1,3-propanediol, THAM, Tris base, Tris(hydroxymethyl)aminomethane, Trometamol, Tris

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H11NO3
CAS Number:
Molecular Weight:
121.14
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

Product Name

Trometamol, European Pharmacopoeia (EP) Reference Standard

grade

pharmaceutical primary standard

API family

trometamol, tromethamine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

NC(CO)(CO)CO

InChI

1S/C4H11NO3/c5-4(1-6,2-7)3-8/h6-8H,1-3,5H2

InChI key

LENZDBCJOHFCAS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets, has been developed and issued under the Authority of the issuing Pharmacopoeia.

For further information and support please go to the website of the issuing Pharmacopoeia.

Application

This European Pharmacopoeia reference standard is intended for use only as specifically prescribed in the European Pharmacopoeia. Their suitability for any other use is not guaranteed and is the sole responsibility of the user. This standard is not intended for human or animal use.
Established for use in the identification of trometamol using Infra-red absorption spectroscopy and preparation of reference solution (a) to test related substances in trometamol using thin-layer chromatography (General text 2.2.27), as per monograph 1053 of European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Trometamol
European Pharmacopoeia Commission and European Directorate for the Quality of Medicines & Healthcare
European pharmacopoeia, 4107-4108 (2017)
Corey A Baron et al.
Magnetic resonance in medicine, 73(3), 1075-1084 (2014-04-12)
An acquisition method that does not increase scan time or specific absorption rate is investigated for reducing the deleterious effects of cerebrospinal fluid (CSF) partial volume effects on diffusion tensor imaging (DTI) tractography. It is based on using a shorter
Filippo Bertoli et al.
Small (Weinheim an der Bergstrasse, Germany), 10(16), 3307-3315 (2014-04-17)
Nanoparticles in contact with cells and living organisms generate quite novel interactions at the interface between the nanoparticle surface and the surrounding biological environment. However, a detailed time resolved molecular level description of the evolving interactions as nanoparticles are internalized
D R Brocks et al.
Clinical pharmacokinetics, 23(6), 415-427 (1992-12-01)
Ketorolac is a new chiral nonsteroidal anti-inflammatory drug (NSAID) which is marketed for analgesia as the racemate. The drug is administered as the water soluble tromethamine salt and is available in tablets or as an intramuscular injection. The absorption of
Xia-Bing Fu et al.
Dalton transactions (Cambridge, England : 2003), 43(23), 8721-8737 (2014-04-29)
Two new water-soluble copper(ii)-dipeptide complexes: [Cu(glygly)(PyTA)]ClO4·1.5H2O (1) and [Cu(glygly)(PzTA)]ClO4·1.5H2O (2) (glygly = glycylglycine anion, PyTA = 2,4-diamino-6-(2'-pyridyl)-1,3,5-triazine and PzTA = 2,4-diamino-6-(2'-pyrazino)-1,3,5-triazine), utilizing two interrelated DNA base-like ligands (PyTA and PzTA), have been synthesized and characterized. The structure elucidation for 1

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service