Skip to Content
Merck
All Photos(1)

Key Documents

ED2SC

Sigma-Aldrich

Ethylenediaminetetraacetic acid calcium disodium salt hydrate

Synonym(s):

Calcium disodium ethylenediaminetetraacetate hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ca(OOCCH2)2NCH2CH2N(CH2COONa)2 · aq
CAS Number:
Molecular Weight:
374.27 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

biological source

synthetic

Quality Level

Assay

97.0-102.0% (titration)

form

powder

reaction suitability

reagent type: chelator

solubility

water: 1.6 g/10 mL, clear, colorless to faintly yellow

storage temp.

room temp

SMILES string

O.[Na+].[Na+].[Ca++].[O-]C(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O

InChI

1S/C10H16N2O8.Ca.2Na.H2O/c13-7(14)3-11(4-8(15)16)1-2-12(5-9(17)18)6-10(19)20;;;;/h1-6H2,(H,13,14)(H,15,16)(H,17,18)(H,19,20);;;;1H2/q;+2;2*+1;/p-4

InChI key

JHECKPXUCKQCSH-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Application

  • MIL-101(Fe) based biomass as permeable reactive barrier applied to EK-PRB remediation of antimony contaminated soil.: The research utilizes EDTA as a chelating agent to enhance the performance of a MIL-101(Fe) based reactive barrier for the electrokinetic remediation of antimony-contaminated soil (Wang et al., 2023).

  • Proteomic signatures of synergistic interactions in antimicrobials.: This proteomics study investigates the synergistic interactions of antimicrobials, highlighting the role of EDTA in enhancing antimicrobial efficacy by disrupting microbial cell membranes (Zhou et al., 2023).

  • Rapid and zero-cost DNA extraction from soft-bodied insects for routine PCR-based applications.: This study presents a novel method for DNA extraction using EDTA, which simplifies and reduces the cost of DNA extraction from soft-bodied insects for PCR applications (Jangra and Ghosh, 2022).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bénédicte Gnangnon et al.
Bio-protocol, 10(11), e3647-e3647 (2021-03-05)
The study of host/pathogen interactions at the cellular level during Plasmodium intra-erythrocytic cycle requires differential extraction techniques aiming to analyze the different compartments of the infected cell. Various protocols have been proposed in the literature to study specific compartments and/or
Matthew A Walker et al.
JCI insight, 6(3) (2021-02-09)
A hallmark of impaired myocardial energetics in failing hearts is the downregulation of the creatine kinase (CK) system. In heart failure patients and animal models, myocardial phosphocreatine content and the flux of the CK reaction are negatively correlated with the
Evaluation of the Trial to Assess Chelation Therapy (TACT): the scientific process, peer review, and editorial scrutiny.
Howard Bauchner et al.
JAMA, 309(12), 1291-1292 (2013-03-28)
Chelation therapy and cardiovascular outcomes.
Bruce Kleinman
JAMA, 310(4), 431-431 (2013-08-07)
Gervasio A Lamas et al.
JAMA, 309(12), 1241-1250 (2013-03-28)
Chelation therapy with disodium EDTA has been used for more than 50 years to treat atherosclerosis without proof of efficacy. To determine if an EDTA-based chelation regimen reduces cardiovascular events. Double-blind, placebo-controlled, 2 × 2 factorial randomized trial enrolling 1708

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service