P1458
Polyethylene glycol solution
40 % (w/w) in H2O, average mol wt 8,000
Synonym(s):
PEG solution
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
CAS Number:
MDL number:
UNSPSC Code:
12352104
PubChem Substance ID:
NACRES:
NA.21
Recommended Products
sterility
0.2 μm filtered
Quality Level
mol wt
average mol wt 8,000
concentration
40 % (w/w) in H2O
storage temp.
2-8°C
SMILES string
C(CO)O
InChI
1S/C2H6O2/c3-1-2-4/h3-4H,1-2H2
InChI key
LYCAIKOWRPUZTN-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
For making aqueous two-phase partitions for cell fractionation
PEG is used as a fusogen to obtain hybridomas for monoclonal antibody production. Induces cell hybridization.
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Liudi Tang et al.
PLoS pathogens, 15(4), e1007742-e1007742 (2019-04-27)
Persistent hepatitis B virus (HBV) infection relies on the establishment and maintenance of covalently closed circular (ccc) DNA, a 3.2 kb episome that serves as a viral transcription template, in the nucleus of an infected hepatocyte. Although evidence suggests that
Sevdan Yılmaz et al.
Fish & shellfish immunology, 78, 140-157 (2018-04-24)
The present study investigated the effects of dietary trans-cinnamic acid (CA) on pre- and post-challenge haematological, serum biochemical, non-specific immune and head kidney gene expression responses of rainbow trout, Oncorhynchus mykiss juveniles. In this regard, fish with an average weight
Sergei A Filichkin et al.
Genome research, 20(1), 45-58 (2009-10-28)
Alternative splicing can enhance transcriptome plasticity and proteome diversity. In plants, alternative splicing can be manifested at different developmental stages, and is frequently associated with specific tissue types or environmental conditions such as abiotic stress. We mapped the Arabidopsis transcriptome
Seyeon Bae et al.
Immune network, 11(3), 175-181 (2011-08-24)
CM1 (centrocyte/-blast marker 1) was defined by a mAb against concanavalin A (Con A) activated PBMC. It is expressed in germinal center of human tonsil and on the surface of activated PBMC as well as cancer cells. Recently, increased productions
Sondra T Bland et al.
Brain, behavior, and immunity, 23(4), 492-497 (2009-06-03)
Glial activation has recently been discovered to modulate several effects of morphine, including analgesia, tolerance, and dependence. The present studies extend this line of investigation by exploring whether glial activation may also affect extracellular levels of dopamine (DA) in the
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service