Skip to Content
Merck
All Photos(2)

Documents

32533

Supelco

Vedaprofen

VETRANAL®, analytical standard

Synonym(s):

(±)-2-(4-Cyclohexylnaphthalen-1-yl)propionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H22O2
CAS Number:
Molecular Weight:
282.38
Beilstein:
5568831
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CC(C(O)=O)c1ccc(C2CCCCC2)c3ccccc13

InChI

1S/C19H22O2/c1-13(19(20)21)15-11-12-16(14-7-3-2-4-8-14)18-10-6-5-9-17(15)18/h5-6,9-14H,2-4,7-8H2,1H3,(H,20,21)

InChI key

VZUGVMQFWFVFBX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Vedaprofen is an arylpropionic acid derivative used as a non-steroidal anti-inflammatory drug (NSAID) in veterinary medicine.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Vedaprofen may be used as an analytical reference standard for the determination of the analyte in:
  • Milk samples by rapid resolution liquid chromatography-tandem mass spectrometry (RRLC-MS/MS).
  • Urine samples by solid-phase extraction (SPE) and LC-MS.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rapid confirmatory analysis of non-steroidal anti-inflammatory drugs in bovine milk by rapid resolution liquid chromatography tandem mass spectrometry.
Dowling G, et al.
Journal of Chromatography A, 1216(46), 8117-8131 (2009)
Comprehensive screening of anabolic steroids, corticosteroids, and acidic drugs in horse urine by solid-phase extraction and liquid chromatography-mass spectrometry.
Ho ENM, et al.
Journal of Chromatography A, 1120(1-2), 38-53 (2006)
Confirmatory analysis of non-steroidal anti-inflammatory drugs in bovine milk by high-performance liquid chromatography with fluorescence detection.
Gallo P, et al.
Journal of Chromatography A, 1217(17), 2832-2839 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service