Skip to Content
Merck
All Photos(3)

Key Documents

O5015

Sigma-Aldrich

Oxybutynin chloride

meets EP, USP testing specifications

Synonym(s):

α-Phenylcyclohexaneglycolic acid 4-(diethylamino)-2-butynyl ester hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H31NO3 · HCl
CAS Number:
Molecular Weight:
393.95
EC Number:
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:
NACRES:
NA.21

Agency

USP/NF
meets EP testing specifications
meets USP testing specifications

Quality Level

form

solid

application(s)

pharmaceutical (small molecule)

storage temp.

room temp

SMILES string

Cl.CCN(CC)CC#CCOC(=O)C(O)(C1CCCCC1)c2ccccc2

InChI

1S/C22H31NO3.ClH/c1-3-23(4-2)17-11-12-18-26-21(24)22(25,19-13-7-5-8-14-19)20-15-9-6-10-16-20;/h5,7-8,13-14,20,25H,3-4,6,9-10,15-18H2,1-2H3;1H

InChI key

SWIJYDAEGSIQPZ-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Muscarinic acetylcholine receptor antagonist.
Muscarinic acetylcholine receptor antagonist; inhibits proliferation of bladder smooth muscle cells, perhaps by downregulation of growth promoting genes.

related product

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Theodore M Johnson et al.
BJU international, 112(1), 100-108 (2013-03-02)
WHAT'S KNOWN ON THE SUBJECT? AND WHAT DOES THE STUDY ADD?: Nocturia is a common and bothersome lower urinary tract symptom, particularly in men. Many single drug therapies have limited benefit. For men who have persistent nocturia despite alpha-blocker therapy
V Schuler et al.
Phytomedicine : international journal of phytotherapy and phytopharmacology, 19(10), 947-951 (2012-07-25)
A broad spectrum of synthetic agents is available for the treatment of overactive bladder. Anti-cholinergic drugs show a poor compliance due to side effects. There is an increasing use of plant extracts in medicine. We have therefore investigated the inhibitory
Jessica T Casey et al.
The Journal of urology, 188(4 Suppl), 1516-1520 (2012-08-23)
Abnormal bladder function following posterior urethral valve ablation can lead to deleterious effects on renal function and urinary continence. We performed a pilot study to determine if bladder dysfunction could be ameliorated by the early administration of oxybutynin. We enrolled
Michael H Johnson et al.
The Journal of urology, 189(4), 1268-1274 (2012-11-06)
Intravesical bacillus Calmette-Guérin is used to decrease recurrence rates of nonmuscle invasive urothelial carcinoma. Irritative urinary symptoms are a common side effect of treatment and frequently limit treatment tolerance. While anticholinergic medications may be used for symptom prophylaxis, to our
Gary G Kay et al.
Clinical drug investigation, 32(10), 707-714 (2012-08-23)
Oxybutynin is a common antimuscarinic therapy for overactive bladder. Transdermally administered oxybutynin chloride topical gel 10% (OTG) has a low propensity for anticholinergic adverse effects and possibly also a low risk of cognitive impairment. A randomized, double-blind, placebo- and active-controlled

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service